Skip to Content
Merck
All Photos(5)

Key Documents

185361

Sigma-Aldrich

Terephthalic acid

greener alternative

98%

Synonym(s):

Benzene-1,4-dicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,4-(CO2H)2
CAS Number:
Molecular Weight:
166.13
Beilstein:
1909333
EC Number:
MDL number:
UNSPSC Code:
12162002
eCl@ss:
39024105
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

98%

form

powder

autoignition temp.

925 °F

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

>300 °C (lit.)

solubility

water: ~0.017 g/L at 25 °C

density

1.58 g/cm3 at 25 °C

greener alternative category

SMILES string

OC(=O)c1ccc(cc1)C(O)=O

InChI

1S/C8H6O4/c9-7(10)5-1-2-6(4-3-5)8(11)12/h1-4H,(H,9,10)(H,11,12)

InChI key

KKEYFWRCBNTPAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Terephthalic acid belongs to the class of monomers known as aromatic dicarboxylic acids. It is primarily used as a key monomer in the production of a high-performance polymer known as polyethylene terephthalate (PET). It is also used to prepare other polymers such as polybutylene terephthalate (PBT), polymer blends, and alloys. Terephthalic acid-based polymers are widely used in various industries including, packaging, textile fibers, polyester resins, polyurethane coatings, polyurethane foams, protective coatings, electrical components, and automotive applications due to their excellent properties such as high thermal stability, chemical resistance, lightweight, transparency, high strength, and durability. In the medical industry, terephthalic acid-based polymers may be used in the production of medical devices and equipment such as surgical sutures, tissue engineering scaffolds, and vascular grafts due to their biocompatibility, chemical resistance, and ease of processing.[1][2][3][4]

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Terephthalic acid can be used:
  • As a monomer in the synthesis of poly(butylene terephthalate) (PBT), a type of polyester, that is used in various fields including Automotive components, textile Industry, packaging materials, electrical and electronic components.
  • As an organic ligand in the synthesis of the cobalt(II) metal–organic framework (MOFs), which finds applications in electrochemical energy storage, catalysis, optoelectronics, and water treatment.
  • Terephthalic acid (TPA) can be synthesized from bio-based materials for a variety of applications, which include the production of polyester fiber, non-fiber field, PET bottles, synthetic perfumes and medicines.
  • Terephthalic acid is used as a linker molecule in the preparation of metal-organic frameworks (MOFs).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Photocatalytic degradation of terephthalic acid using titania and zinc oxide photocatalysts: Comparative study
Shafaei A, et al.
Desalination, 252(1-3), 8-16 (2010)
Synthesis of ethylene glycol and terephthalic acid from biomass for producing PET
Pang J, et al.
Green Chemistry, 18(2), 342-359 (2016)
Biobased terephthalic acid technologies: a literature review
Collias DI, et al.
Industrial Biotechnology (New Rochelle, N.Y.), 10(2), 91-105 (2014)
Jingqi Tian et al.
Biosensors & bioelectronics, 71, 1-6 (2015-04-17)
Considerable recent attention has been paid to homogeneous fluorescent DNA detection with the use of nanostructures as a universal "quencher", but it still remains a great challenge to develop such nanosensor with the benefits of low cost, high speed, sensitivity
Akram Hijazi et al.
Dalton transactions (Cambridge, England : 2003), 42(14), 4848-4858 (2013-02-01)
We report the syntheses and characterizations, in the solid state and in solution, of three new cyclic polyoxothiomolybdates self-assembled around 2,5-dimethylterephthalate (DMT) and 2,5-bis(trifluoromethyl)terephthalate (DFMT) ligands, namely [Mo12DMT](2-), [Mo12DFMT](2-) and [Mo16DFMT](2-). A series of these two Mo12-compounds completed by the

Articles

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Is this product a powder or in pellets? Is this product a powder or pellets? 185361 Sigma-Aldrich Terephthalic acid

    1 answer
    1. This product is offered in powdered format as listed in the 'PROPERTIES' section.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service