Merck
  • Liver X receptor regulates mouse GM-CSF-derived dendritic cell differentiation in vitro.
Liver X receptor regulates mouse GM-CSF-derived dendritic cell differentiation in vitro.
Molecular immunology (2014-04-22)
Limei Zhong, Quan Yang, Wen Xie, Jie Zhou
ABSTRACT
Liver X receptors (LXRs) are nuclear receptors that play an essential role in lipid and cholesterol metabolism. Emerging studies indicate a potential function for LXRs in regulating dendritic cell (DC)-dependent immune responses; however, the role of LXRs in DC differentiation is largely unknown. Here, we report that LXRα regulates the differentiation of mouse GM-CSF-derived DCs. Activation or overexpression of LXRα significantly enhanced myeloid DC differentiation from mouse bone marrow (BM) cells, while siRNA-mediated knockdown of LXRα suppressed DC differentiation. In addition, we demonstrated that LXR agonist-programmed DCs showed an increased capacity for stimulating T-cell proliferation. Mechanistic studies showed that activation of LXR could inhibit the phosphorylation of STAT3 and downregulate the expression of its target, S100A9, an important negative regulator of myeloid DC differentiation. We also found that Histone deacetylase (HDAC) inhibition interfered with the effect of LXR on STAT3 signaling via acetylation of STAT3. Chromatin immunoprecipitation assays confirmed that LXR activation and HDAC inhibition balanced the recruitment of STAT3 to the S100A9 promoter, which involved distinct post-translational modifications of STAT3. In conclusion, our observations demonstrated a novel role for LXRα in GM-CSF-derived DC differentiation and revealed the underlying mechanism.
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
6-Carboxyfluorescein diacetate, ≥95% (HPLC)
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Cucurbitacin I hydrate, ≥95% (HPLC), solid
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Nr1h3
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Supelco
Water, for TOC analysis
Sigma-Aldrich
MISSION® esiRNA, targeting human NR1H2
Supelco
Water, suitable for ion chromatography
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, ACS reagent, for ultratrace analysis
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, suitable for HPLC