Skip to Content
Merck
All Photos(2)

Documents

W1754

Sigma-Aldrich

Water

PCR Reagent, suitable for PCR

Synonym(s):

H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
EC Number:
MDL number:
UNSPSC Code:
12191602
PubChem Substance ID:
NACRES:
NA.25

product name

Water, PCR Reagent

grade

PCR Reagent

vapor density

<1 (vs air)

vapor pressure

3 mmHg

sterility

sterile-filtered

form

liquid

packaging

vial of 1.5 mL

technique(s)

PCR: suitable

refractive index

n20/D 1.34 (lit.)

pH

5-7

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

foreign activity

DNase, none detected
RNase, none detected

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

PCR grade water is sterile-filtered. It is free from exonucleases (DNAse, RNAse) and endonuclease (NICKase) and is also free from nucleic acid contamination.

Application

Water has been used:

  • as a component of the reaction mixture and as a diluting agent in microfluidic RT-qPCR
  • as a component of the reaction mixture for the amplification of products from fungal (Trametes versicolor) DNA
  • as a diluting agent and as a component of the reaction mixture for the amplification of cDNA
  • Water has been used to make up the final volume of the sample in polymerase chain reaction (PCR)

Suitability

Suitable for polymerase chain reaction (PCR)

Other Notes

Easily compare specifications for Water products with the Water specification table.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francine Marciano-Cabral et al.
Applied and environmental microbiology, 69(10), 5864-5869 (2003-10-09)
The free-living amoeboflagellate Naegleria fowleri is the causative agent of primary amoebic meningoencephalitis (PAM), a rapidly fatal disease of the central nervous system. In the United States, the disease is generally acquired while swimming and diving in freshwater lakes and
Incidence and survival of non-O157 verocytotoxigenic Escherichia coli in soil
Bolton DJ, et al.
Journal of Applied Microbiology, 111(2), 484-490 (2011)
Multiplex PCR for rapid detection of genes encoding acquired metallo-beta-lactamases.
Matthew J Ellington et al.
The Journal of antimicrobial chemotherapy, 59(2), 321-322 (2006-12-23)
Fatty acid secretion by the white-rot fungus, Trametes versicolor
Guyu H, et al.
Journal of Industrial and Engineering Chemistry, 49(1) (2022)
If you?ve seen one worm, have you seen them all? Spatial, community, and genetic variability of tubificid communities in Montana
Lodh N, et al.
Freshwater science, 34(3), 909-917 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service