Skip to Content
Merck
All Photos(1)

Key Documents

95289

Sigma-Aldrich

Water

for cell biology

Synonym(s):

Water for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.71
grade:
for cell biology
application(s):
cell analysis
bp:
100 °C (lit.)

Product Name

Water, for cell biology, sterile ultrafiltered

grade

for cell biology

Quality Level

form

liquid

quality

sterile ultrafiltered

shelf life

limited shelf life, expiry date on the label

impurities

mycoplasma, none detected (fluorimetric detection of DNA traces; Hoechst test with Bisbenzimide H 33258 (Cat. No 14530))
≤0.005 EU/mL endoxins

refractive index

n20/D 1.34 (lit.)

pH

5-7

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

cation traces

Ca: <10 ppb
Mg: <10 ppb

application(s)

cell analysis

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Water has been used:
  • to rinse the corroded and noncorroded disks in lipopolysaccharide affinity assay
  • during hydrogel synthesis and preparation
  • in the preparation of 2-mercaptoethanol and CpG-ODN stock solution
  • in the preparation of lipopolysaccharides (LPS)

Other Notes

Easily compare specifications for Water products with the Water specification table.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Comparative analysis of QCM and SPR techniques for the optimization of immobilization sequences
Ansorena P, et al.
Sensors and Actuators B, Chemical, 155(2), 667-672 (2011)
Influence of corrosion on lipopolysaccharide affinity for two different titanium materials
Barao VAR, et al.
The Journal of Prosthetic Dentistry, 110(6), 462-470 (2013)
Biomimetic poly (amidoamine) hydrogels as synthetic materials for cell culture
Jacchetti E, et al.
Journal of Nanobiotechnology, 6(1), 14-14 (2008)
Production and analysis of immunomodulatory excretory-secretory products from the mouse gastrointestinal nematode Heligmosomoides polygyrus bakeri
Valanparambil RM, et al.
Nature Protocols, 9(12), 2740-2740 (2014)
Philippe Robert et al.
Investigative radiology, 50(8), 473-480 (2015-06-25)
To prospectively compare in healthy rats the effect of multiple injections of macrocyclic (gadoterate meglumine) and linear (gadodiamide) gadolinium-based contrast agents (GBCAs) on T1-weighted signal intensity in the deep cerebellar nuclei (DCN), including the dentate nucleus. Healthy rats (n =

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service