Skip to Content
Merck
All Photos(2)

Key Documents

W3513

Sigma-Aldrich

Water

HPLC grade, suitable for HPLC

Synonym(s):

H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
EC Number:
MDL number:
UNSPSC Code:
12191602
PubChem Substance ID:
NACRES:
NA.25
grade:
BioPerformance Certified
technique(s):
cell culture | mammalian: suitable
bp:
100 °C (lit.)
vapor pressure:
3 mmHg
Pricing and availability is not currently available.

Product Name

Water, BioPerformance Certified

grade

BioPerformance Certified

vapor density

<1 (vs air)

vapor pressure

3 mmHg

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

refractive index

n20/D 1.34 (lit.)

pH

5-7

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

absorption

≤0.05 at 290

foreign activity

DNase, RNase, none detected

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Water has been used:
  • as a component of the reaction mixture for polymerase chain reaction (PCR)[1][2]
  • in the dilution of plasmids for real-time PCR[3]
  • as a component of 1X TE buffer and library elution buffer (LEB)[4]

Preparation Note

This product is prepared from double distilled and deionized water

Other Notes

Easily compare specifications for Water products with the Water specification table.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biochemical kinetics of fermentative hydrogen production by Clostridium butyricum W5
Wang X, et al.
International Journal of Hydrogen Energy, 34(2), 791-798 (2009)
Franziska Greulich et al.
STAR protocols, 2(3), 100609-100609 (2021-07-01)
Quantifying differential genome occupancy by chromatin immunoprecipitation (ChIP) remains challenging due to variation in chromatin fragmentation, immunoprecipitation efficiencies, and intertube variability. In this protocol, we add heterologous spike-ins from Drosophila chromatin as an internal control to the mice chromatin before
Philippe Robert et al.
Investigative radiology, 50(8), 473-480 (2015-06-25)
To prospectively compare in healthy rats the effect of multiple injections of macrocyclic (gadoterate meglumine) and linear (gadodiamide) gadolinium-based contrast agents (GBCAs) on T1-weighted signal intensity in the deep cerebellar nuclei (DCN), including the dentate nucleus. Healthy rats (n =
Susu Duan et al.
Nature communications, 5, 5029-5029 (2014-10-10)
Oseltamivir-resistant H1N1 influenza viruses carrying the H275Y neuraminidase mutation predominated worldwide during the 2007-2009 seasons. Although several neuraminidase substitutions were found to be necessary to counteract the adverse effects of H275Y, the order and impact of evolutionary events involved remain
Mary A Rodgers et al.
The Journal of experimental medicine, 211(7), 1333-1347 (2014-06-25)
Linear ubiquitination is a newly discovered posttranslational modification that is currently restricted to a small number of known protein substrates. The linear ubiquitination assembly complex (LUBAC), consisting of HOIL-1L, HOIP, and Sharpin, has been reported to activate NF-κB-mediated transcription in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service