Skip to Content
Merck
All Photos(2)

Key Documents

320072

Sigma-Aldrich

Water

ACS reagent, suitable for EPA 601

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
EC Number:
MDL number:
UNSPSC Code:
12191602
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
bp:
100 °C (lit.)
vapor pressure:
3 mmHg
Pricing and availability is not currently available.

Product Name

Water, ACS reagent

grade

ACS reagent

Quality Level

Agency

suitable for EPA 601

vapor density

<1 (vs air)

vapor pressure

3 mmHg

form

liquid

impurities

MnO4- reducers, passes test
≤0.01% Silicate (as SiO2)
≤0.01 ppm silicate

refractive index

n20/D 1.34 (lit.)

pH

6.0-8.0 (25 °C)

conductivity

≤2 μmho per cm at 25 °C

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

anion traces

chloride (Cl-): ≤0.4 ppm
nitrate (NO3-): ≤0.4 ppm
phosphate (PO43-): ≤1.0 ppm
sulfate (SO42-): ≤1.0 ppm

cation traces

heavy metals (as Pb): ≤0.01 ppm

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Water can be used as a:
  • Solvent to ensure the appropriate deposition and recrystallization of the matrix material used in the matrix-assisted laser desorption/ionization (MALDI) analysis.[1]
  • Mobile phase component in the high-performance liquid chromatography (HPLC) method used for the separation of polar compounds.[2]

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Matrix vapor deposition/recrystallization and dedicated spray preparation for high-resolution scanning microprobe matrix-assisted laser desorption/ionization imaging mass spectrometry (SMALDI-MS) of tissue and single cells
Bouschen W, et al.
Rapid Communications in Mass Spectrometry, 24(3), 355-364 (2010)
Evaluation of various HILIC materials for the fast separation of polar compounds
Chauve B, et al.},
Journal of Separation Science, 33(6-7), 752-764 (2010)
Manolis Mandalakis et al.
Journal of chromatography. A, 1217(1), 143-150 (2009-12-08)
The analysis of amino acids by gas chromatography mass spectrometry (GC-MS) after their derivatization with N-(t-butyldimethylsilyl)-N-methyltrifluoroacetamide was investigated as an alternative approach for the determination of free (FAA) and combined amino acids (CAA) in aerosols. This technique showed excellent linearity
S Bekesova et al.
Journal of proteomics, 75(7), 2216-2224 (2012-02-14)
N-glycosylation of proteins provides a rich source of information on liver disease progression because majority of serum glycoproteins, with the exception of immunoglobulins, are secreted by the liver. In this report, we present results of an optimized workflow for MALDI-TOF
Martin Fischnaller et al.
Analytica chimica acta, 761, 92-101 (2013-01-15)
Phosphorylation of proteins is an important cellular regulatory process. The analysis of protein phosphorylation is challenging due to the high dynamic range and low abundance natures of phosphorylated species. Mass spectrometry (MS) of phosphopeptides obtained from tryptic protein digests is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service