W4502
Water
suitable for electrophoresis, PCR
Synonym(s):
DNase-Free Water, Not DEPC Treated, Nuclease-Free Water, Protease-Free Water, RNase-Free Water
About This Item
Recommended Products
product name
Water, Nuclease-Free Water, for Molecular Biology
vapor density
<1 (vs air)
Quality Level
vapor pressure
3 mmHg
sterility
0.1 μm filtered
form
liquid
technique(s)
PCR: suitable
electrophoresis: suitable
refractive index
n20/D 1.34 (lit.)
pH
5-7
bp
100 °C (lit.)
mp
0 °C (lit.)
density
1.000 g/mL at 3.98 °C (lit.)
foreign activity
DNase, RNase, Protease, free
SMILES string
O
InChI
1S/H2O/h1H2
InChI key
XLYOFNOQVPJJNP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- in cDNA synthesis and PCR amplification
- for the dilution of rumen DNA sample prior to qPCR assay
- in PCR (polymerase chain reaction) and agarose gel electrophoresis
- for the elution of RNA during RNA isolation
Features and Benefits
- Nuclease-Free Water
- DNase-Free, RNase-Free, Protease- Free
- Not DEPC Treated
- 0.1 um Filtered
- Prepared from Double Distilled and Deionized Water
- Suitable for use in Molecular Applications
This product is 0.1um filtered. It is designated as a Molecular Biology Reagent and is suitable for molecular biology applications. This product has been analyzed for the absence of nucleases and proteases, and has undergone bioburden analysis. It has not been treated with diethyl pyrocarbonate (DEPC).
Preparation Note
Other Notes
related product
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service