Skip to Content
Merck
All Photos(1)

Documents

W3500

Sigma-Aldrich

Water

sterile-filtered, BioReagent, suitable for cell culture

Synonym(s):

Cell culture grade water, Ultra-filtered Water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

vapor density

<1 (vs air)

Quality Level

vapor pressure

3 mmHg

sterility

sterile-filtered

product line

BioReagent

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

≤1 EU/mL endotoxin

refractive index

n20/D 1.34 (lit.)

pH

5-7

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is a very low endotoxin (less than 1 EU/mL), 0.2 micron filtered.

Application

For use in the preparation of cell culture media, and cell suspension and washing solutions.

Other Notes

Easily compare specifications for Water products with the Water specification table

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Philippe Robert et al.
Investigative radiology, 50(8), 473-480 (2015-06-25)
To prospectively compare in healthy rats the effect of multiple injections of macrocyclic (gadoterate meglumine) and linear (gadodiamide) gadolinium-based contrast agents (GBCAs) on T1-weighted signal intensity in the deep cerebellar nuclei (DCN), including the dentate nucleus. Healthy rats (n =
Susu Duan et al.
Nature communications, 5, 5029-5029 (2014-10-10)
Oseltamivir-resistant H1N1 influenza viruses carrying the H275Y neuraminidase mutation predominated worldwide during the 2007-2009 seasons. Although several neuraminidase substitutions were found to be necessary to counteract the adverse effects of H275Y, the order and impact of evolutionary events involved remain
Mary A Rodgers et al.
The Journal of experimental medicine, 211(7), 1333-1347 (2014-06-25)
Linear ubiquitination is a newly discovered posttranslational modification that is currently restricted to a small number of known protein substrates. The linear ubiquitination assembly complex (LUBAC), consisting of HOIL-1L, HOIP, and Sharpin, has been reported to activate NF-κB-mediated transcription in
Yasushi Itoh et al.
PLoS pathogens, 10(6), e1004192-e1004192 (2014-06-20)
Highly pathogenic avian influenza (HPAI) viruses of the H5N1 subtype often cause severe pneumonia and multiple organ failure in humans, with reported case fatality rates of more than 60%. To develop a clinical antibody therapy, we generated a human-mouse chimeric
M Grusdat et al.
Cell death and differentiation, 21(7), 1050-1060 (2014-02-18)
CD8(+) T-cell functions are critical for preventing chronic viral infections by eliminating infected cells. For healthy immune responses, beneficial destruction of infected cells must be balanced against immunopathology resulting from collateral damage to tissues. These processes are regulated by factors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service