Merck
  • Synthesis of empagliflozin, a novel and selective sodium-glucose co-transporter-2 inhibitor, labeled with carbon-14 and carbon-13.
Synthesis of empagliflozin, a novel and selective sodium-glucose co-transporter-2 inhibitor, labeled with carbon-14 and carbon-13.
Journal of labelled compounds & radiopharmaceuticals (2014-10-22)
Matt Hrapchak, Bachir Latli, Xiao-Jun Wang, Heewon Lee, Scot Campbell, Jinhua J Song, Chris H Senanayake
ABSTRACT
Empagliflozin, (2S,3R,4R,5S,6R)-2-[4-chloro-3-[[4-[(3S)-oxolan-3-yl]oxyphenyl]methyl]phenyl]-6-(hydroxymethyl)oxane-3,4,5-triol was recently approved by the FDA for the treatment of chronic type 2 diabetes mellitus. Herein, we report the synthesis of carbon-13 and carbon-14 labeled empagliflozin. Carbon-13 labeled empagliflozin was prepared in five steps and in 34% overall chemical yield starting from the commercially available α-D-glucose-[(13)C6]. For the radiosynthesis, the carbon-14 atom was introduced in three different positions of the molecule. In the first synthesis, Carbon-14 D-(+)-gluconic acid δ-lactone was used to prepare specifically labeled empagliflozin in carbon-1 of the sugar moiety in four steps and in 19% overall radiochemical yield. Carbon-14 labeled empagliflozin with the radioactive atom in the benzylic position was obtained in eight steps and in 7% overall radiochemical yield. In the last synthesis carbon-14 uniformly labeled phenol was used to give [(14)C]empagliflozin in eight steps and in 18% overall radiochemical yield. In all these radiosyntheses, the specific activities of the final compounds were higher than 53 mCi/mmol, and the radiochemical purities were above 98.5%.
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
D-Gluconic acid solution, 49-53 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)