W200611
Acetic acid
≥99.5%, FCC, FG
Synonym(s):
Glacial acetic acid
About This Item
Fragrance grade
Halal
Kosher
Recommended Products
biological source
synthetic
Quality Level
300
400
grade
FG
Fragrance grade
Halal
Kosher
Agency
follows IFRA guidelines
meets purity specifications of JECFA
reg. compliance
EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.814
FDA 21 CFR 173.370
FDA 21 CFR 184.1005
vapor density
2.07 (vs air)
vapor pressure
15.2 hPa ( 20.0 °C)
73.3 hPa ( 50.0 °C)
Assay
≥99.5%
form
liquid
autoignition temp.
800 °F
shelf life
Expiration date period - 5 years
expl. lim.
16 %, 92 °F
4 %, 59 °F
color
colorless
refractive index
n20/D 1.371 (lit.)
pH
2.5 (20 °C, 50 g/L)
bp
117-118 °C (lit.)
mp
16.2 °C (lit.)
transition temp
solidification point ≥15.6 °C
density
1.04 g/mL at 25 °C (lit.)
cation traces
As: ≤3 ppm
Cd: ≤1 ppm
Hg: ≤1 ppm
heavy metals (as Pb): ≤0.5 ppm
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
SMILES string
[F2C(F2C)13F3C]C(O)=O
food allergen
no known allergens
fragrance allergen
no known allergens
Organoleptic
sour
InChI
1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)
InChI key
QTBSBXVTEAMEQO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
102.2 °F - closed cup
Flash Point(C)
39 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service