Skip to Content
Merck
All Photos(1)

Key Documents

45726

Sigma-Aldrich

Acetic acid

for luminescence, BioUltra, ≥99.5% (GC)

Synonym(s):

Glacial acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
Beilstein:
506007
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

grade

for luminescence

vapor density

2.07 (vs air)

vapor pressure

15.2 hPa ( 20.0 °C)
73.3 hPa ( 50.0 °C)

product line

BioUltra

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

800 °F

expl. lim.

16 %, 92 °F
4 %, 59 °F

impurities

insoluble matter, passes filter test

evapn. residue

≤0.0005%

color

colorless

refractive index

n20/D 1.371 (lit.)

pH

2.5 (20 °C, 50 g/L)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.5 mg/kg
phosphate (PO43-): ≤0.5 mg/kg
sulfate (SO42-): ≤0.5 mg/kg

cation traces

Ag: ≤0.01 mg/kg
Al: ≤0.05 mg/kg
As: ≤0.01 mg/kg
Ba: ≤0.01 mg/kg
Be: ≤0.01 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.2 mg/kg
Cd: ≤0.02 mg/kg
Co: ≤0.01 mg/kg
Cr: ≤0.05 mg/kg
Cu: ≤0.01 mg/kg
Fe: ≤0.2 mg/kg
Ge: ≤0.05 mg/kg
K: ≤0.1 mg/kg
Li: ≤0.01 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.01 mg/kg
Mo: ≤0.02 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.05 mg/kg
Pb: ≤0.02 mg/kg
Sr: ≤0.01 mg/kg
Ti: ≤0.01 mg/kg
Tl: ≤0.05 mg/kg
V: ≤0.01 mg/kg
Zn: ≤0.05 mg/kg
Zr: ≤0.1 mg/kg

SMILES string

[F2C(F2C)13F3C]C(O)=O

λ

neat

UV absorption

λ: 260 nm Amax: ≤0.1
λ: 280 nm Amax: ≤0.02

suitability

corresponds for luminescence
suitable for luminescence

Organoleptic

pungent

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katharine Trenholme et al.
Antimicrobial agents and chemotherapy, 58(7), 3666-3678 (2014-04-16)
Therapies to prevent transmission of malaria parasites to the mosquito vector are a vital part of the global malaria elimination agenda. Primaquine is currently the only drug with such activity; however, its use is limited by side effects. The development
Takashi Taniguchi et al.
Arthritis & rheumatology (Hoboken, N.J.), 67(2), 517-526 (2014-11-12)
Fli-1, a potential predisposing factor for systemic sclerosis (SSc), is constitutively down-regulated in the lesional skin of patients with SSc by an epigenetic mechanism. To investigate the impact of Fli-1 deficiency on the induction of an SSc phenotype in various
Isabelle Brunet et al.
The Journal of clinical investigation, 124(7), 3230-3240 (2014-06-18)
Autonomic sympathetic nerves innervate peripheral resistance arteries, thereby regulating vascular tone and controlling blood supply to organs. Despite the fundamental importance of blood flow control, how sympathetic arterial innervation develops remains largely unknown. Here, we identified the axon guidance cue
Xianbao Deng et al.
The New phytologist, 201(4), 1469-1483 (2013-11-26)
• Chalcone synthase (CHS) is the key enzyme in the first committed step of the flavonoid biosynthetic pathway and catalyzes the stepwise condensation of 4-coumaroyl-CoA and malonyl-CoA to naringenin chalcone. In plants, CHS is often encoded by a small family
Uli S Herrmann et al.
Science translational medicine, 7(299), 299ra123-299ra123 (2015-08-08)
Prions cause transmissible spongiform encephalopathies for which no treatment exists. Prions consist of PrP(Sc), a misfolded and aggregated form of the cellular prion protein (PrP(C)). We explore the antiprion properties of luminescent conjugated polythiophenes (LCPs) that bind and stabilize ordered

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service