Skip to Content
Merck
All Photos(1)

Key Documents

48688

Supelco

Phenol solution

certified reference material, 500 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O
CAS Number:
Molecular Weight:
94.11
Beilstein:
6428463
EC Number:
MDL number:
UNSPSC Code:
12000000
eCl@ss:
39023301
PubChem Substance ID:
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Agency

EPA TO-8
NIOSH 2001
OSHA 32

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

500 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-30°C

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Looking for similar products? Visit Product Comparison Guide

General description

Phenol can be obtained via a two-step process involving the oxidation of toluene by atmospheric oxygen, in the presence of cobalt catalyst to form benzoic acid as the major product, which is followed by the decarboxylation of benzoic acid in the presence of air and copper catalyst.[1] It mostly serves as an important chemical intermediate in the manufacture of synthetic organic materials.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Gardziella.A, et al.
Phenolic Resins: Chemistry, Applications, Standardization, Safety and Ecology (2000)
Neff.M.J
Bioaccumulation in Marine Organisms: Effect of Contaminants from Oil Well Produced Water (2002)
Masaharu Nakamura et al.
Bioorganic & medicinal chemistry, 21(7), 1643-1651 (2013-03-07)
We previously reported that bis-phenol derivatives, including LG190178 (3a), possess not only vitamin D receptor (VDR) agonistic activity, but also androgen receptor (AR) antagonistic activity. Here, we describe the design, synthesis and evaluation of silicon-containing bis-phenol derivatives, with the objective
Rita A Busuttil et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(10), 2761-2772 (2014-03-25)
Gene-expression profiling has revolutionized the way we think about cancer and confers the ability to observe the synchronous expression of thousands of genes. The use of putative genome-level expression profiles has allowed biologists to observe the complex interactions of genes
Mark J Lee et al.
PLoS pathogens, 11(10), e1005187-e1005187 (2015-10-23)
Of the over 250 Aspergillus species, Aspergillus fumigatus accounts for up to 80% of invasive human infections. A. fumigatus produces galactosaminogalactan (GAG), an exopolysaccharide composed of galactose and N-acetyl-galactosamine (GalNAc) that mediates adherence and is required for full virulence. Less

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service