Skip to Content
Merck
All Photos(1)

Key Documents

46344

Supelco

Phenol solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O
CAS Number:
Molecular Weight:
94.11
Beilstein:
6428463
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenol belongs to the class of aromatic volatile organic compounds (VOCs).

Application

Phenol may be used as an analytical reference standard for the determination of phenol in:
  • Air samples from swine and dairy barns by thermal desorption (TD) module coupled with multidimensional gas chromatography (MDGC)-mass spectrometry (MS)/olfactometry (O).
  • Exhaled breath samples from tobacco cigarette and electronic cigarette smokers by GC-MS.
  • Bovine breath samples by solid-phase microextraction (SPME) and GC-MS.
  • Bovine blood samples by headspace in-tube extraction (HS-INTEX) GC-MS.
  • Industrial and urban air samples by TD-GC-MS.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Field air sampling and simultaneous chemical and sensory analysis of livestock odorants with sorbent tubes and GC-MS/olfactometry.
Zhang S, et al.
Sensors and Actuators B, Chemical, 146(2), 427-432 (2010)
A rapid method for the chromatographic analysis of volatile organic compounds in exhaled breath of tobacco cigarette and electronic cigarette smokers.
Marco E and Grimalt JO
Journal of Chromatography A, 1410(2), 51-59 (2015)
Development and validation of a method for air-quality and nuisance odors monitoring of volatile organic compounds using multi-sorbent adsorption and gas chromatography/mass spectrometry thermal desorption system.
Ribes A, et al.
Journal of Chromatography A, 1140(1-2), 44-55 (2007)
Sampling and analysis of volatile organic compounds in bovine breath by solid-phase microextraction and gas chromatography-mass spectrometry.
Spinhirne JP, et al.
Journal of Chromatography A, 1025(1), 63-69 (2004)
Ilpo Rasanen et al.
Journal of analytical toxicology, 34(3), 113-121 (2010-04-22)
A novel headspace in-tube extraction gas chromatography-mass spectrometry (ITEX-GC-MS) approach was developed for broad-scale analysis of low molecular weight organic compounds in blood and/or urine. One sample was analyzed following in-vial derivatization with dimethyl sulfate for ethylene glycol (EG), glycolic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service