Skip to Content
Merck
All Photos(1)

Key Documents

90577

Millipore

Bifido Selective Supplement B

suitable for microbiology

Synonym(s):

Acetic acid solution, Acetic acid Supplement

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
Beilstein:
506007
EC Number:
MDL number:
UNSPSC Code:
41171614
PubChem Substance ID:
NACRES:
NA.85

sterility

sterile

Quality Level

form

liquid

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

microbiology

suitability

Bifidobacterium spp.

SMILES string

CC(O)=O

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

for the selective isolation of Bifidobacterium species from milk products

Compatibility

(per vial, sufficient for 500 ml medium)
Glacial acetic acid (1%) 5 ml

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Taniguchi et al.
Arthritis & rheumatology (Hoboken, N.J.), 67(2), 517-526 (2014-11-12)
Fli-1, a potential predisposing factor for systemic sclerosis (SSc), is constitutively down-regulated in the lesional skin of patients with SSc by an epigenetic mechanism. To investigate the impact of Fli-1 deficiency on the induction of an SSc phenotype in various
Isabelle Brunet et al.
The Journal of clinical investigation, 124(7), 3230-3240 (2014-06-18)
Autonomic sympathetic nerves innervate peripheral resistance arteries, thereby regulating vascular tone and controlling blood supply to organs. Despite the fundamental importance of blood flow control, how sympathetic arterial innervation develops remains largely unknown. Here, we identified the axon guidance cue
Katharine Trenholme et al.
Antimicrobial agents and chemotherapy, 58(7), 3666-3678 (2014-04-16)
Therapies to prevent transmission of malaria parasites to the mosquito vector are a vital part of the global malaria elimination agenda. Primaquine is currently the only drug with such activity; however, its use is limited by side effects. The development
Ilse Cleenwerck et al.
International journal of food microbiology, 125(1), 2-14 (2008-02-02)
Acetic acid bacteria are Gram-negative, obligate aerobic bacteria that have the ability to incompletely oxidize alcohols or sugars to organic acids as end products. They are widespread in nature and most of them are capable to oxidize ethanol as substrate
F Commodari et al.
NMR in biomedicine, 4(4), 192-200 (1991-08-01)
1H NMR was used to analyze human cerebrospinal fluid (CSF) from a group of neurological disease controls and from a vitamin B12 deficient patient. The spectra were acquired at either 7.06 or 9.40 T at ambient temperature with CSF freeze

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service