Skip to Content
Merck
All Photos(3)

Documents

296147

Sigma-Aldrich

Dimethyl sulfoxide-d6

99.9 atom % D, contains 0.03 % (v/v) TMS

Synonym(s):

(Methyl sulfoxide)-d6, DMSO-d6, Hexadeuterodimethyl sulfoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)2SO
CAS Number:
Molecular Weight:
84.17
Beilstein:
1237248
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

0.42 mmHg ( 20 °C)

Quality Level

isotopic purity

99.9 atom % D

Assay

99% (CP)

form

liquid

autoignition temp.

573 °F

contains

0.03 % (v/v) TMS

expl. lim.

42 %

technique(s)

NMR: suitable

impurities

≤0.0250% water
water

refractive index

n20/D 1.476 (lit.)

bp

189 °C (lit.)

mp

20.2 °C (lit.)

density

1.190 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3/i1D3,2D3

InChI key

IAZDPXIOMUYVGZ-WFGJKAKNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl sulfoxide-d6 is a deuterated NMR solvent that is useful in NMR-based research and analyses.

Application


  • Estimation of dissociation constants (pKa′s) of oximes from proton chemical shifts in dimethyl sulfoxide solution: This research utilizes Dimethyl sulfoxide-d₆ (DMSO-d₆) to determine pKa values of oximes, demonstrating its crucial role in enhancing the accuracy of proton NMR spectroscopy for chemical analysis and research within pharmaceutical contexts (Kurtz AP, D′Silva TD, 1987).

  • Proton nuclear magnetic resonance studies on methylthiohydantoins, thiohydantoins, and hydantoins of amino acids: DMSO-d₆ is extensively employed to investigate the structure and behavior of various hydantoin derivatives in amino acid research, proving essential for understanding protein structures and functions, with implications for both academic and pharmaceutical industries (Suzuki T, Tomioka T, Tuzimura K, 1977).

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

190.4 °F

Flash Point(C)

88 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M G Nair et al.
Applied and environmental microbiology, 57(2), 434-439 (1991-02-01)
Two isoflavonoids isolated from clover roots grown under phosphate stress were characterized as formononetin (7-hydroxy,4'-methoxy isoflavone) and biochanin A (5,7-dihydroxy,4'-methoxy isoflavone). At 5 ppm, these compounds stimulated hyphal growth in vitro and root colonization of an undescribed vesicular-arbuscular mycorrhiza, a
Using high-performance quantitative NMR (HP-qNMR?) for certifying traceable and highly accurate purity values of organic reference materials with uncertainties< 0.1%.
Weber M, et al.
Accreditation and Quality Assurance, 18(2), 91-98 (2013)
Diode laser measurements of CD3 quantum yields and internal energy for the dissociation of dimethyl sulfoxide-d6.
Rudolph RN, et al.
J. Chem. Phys. , 106(4), 1346-1352 (1997)
Barbara Parrino et al.
European journal of medicinal chemistry, 94, 367-377 (2015-03-18)
Three new ring systems, pyrido[2',3':3,4]pyrrolo[1,2-a]quinoxalines, pyrido[3',2':3,4]pyrrolo[1,2-a]quinoxalines and pyrido[2',3':5,6]pyrazino[2,1-a]isoindoles, were synthesized through an aza-substitution on the already active isoindolo-quinoxaline system and in particular in the position 7 or 4 of the isoindole moiety and in position 5 of the quinoxaline portion.
Priya P Netalkar et al.
European journal of medicinal chemistry, 79, 47-56 (2014-04-12)
Air and moisture stable coordination compounds of late first row transition metals, viz. Co(II), Ni(II), Cu(II) and Zn(II), with a newly designed ligand, 2-(2-benzo[d]thiazol-2-yl)hydrazono)propan-1-ol (LH), were prepared and successfully characterized using various spectro-analytical techniques. The molecular structures of the ligand

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service