Skip to Content
Merck
All Photos(4)

Key Documents

Z0375

Sigma-Aldrich

trans-Zeatin-riboside

~95%

Synonym(s):

9-(β-D-Ribofuranosyl)-trans-zeatin, N6-(trans-4-Hydroxy-3-methyl-2-buten-1-yl)adenosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21N5O5
CAS Number:
Molecular Weight:
351.36
Beilstein:
5460894
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Assay

~95%

form

powder

solubility

acetic acid: 50 mg/mL, clear, colorless

application(s)

agriculture

storage temp.

−20°C

SMILES string

C\C(CO)=C/CNc1ncnc2n(cnc12)[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O

InChI

1S/C15H21N5O5/c1-8(4-21)2-3-16-13-10-14(18-6-17-13)20(7-19-10)15-12(24)11(23)9(5-22)25-15/h2,6-7,9,11-12,15,21-24H,3-5H2,1H3,(H,16,17,18)/b8-2+/t9-,11-,12-,15-/m1/s1

InChI key

GOSWTRUMMSCNCW-HNNGNKQASA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Koenig RL; Morris RO; Polacco JC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ding Huang et al.
Ying yong sheng tai xue bao = The journal of applied ecology, 17(2), 210-214 (2006-05-19)
The study on the dynamics of soluble sugar and endogenous hormone contents in steppe grass species Elymus sibiricus, E. rectisetus, Leymus chinensis, L. racemosu and Stipa krylovii during their germination period in spring showed that high content GA and low
Lydia B Vysotskaya et al.
Journal of experimental botany, 61(13), 3709-3717 (2010-07-21)
Heat shock (HS) at 40 degrees C was given to the root system of Nicotiana tabacum wild type (WT) and to HSIPT transgenic plants transformed with the bacterial cytokinin biosynthesis gene isopentenyltransferase (ipt) cloned behind the heat shock 70 promoter
Marta Oñate et al.
Annals of botany, 104(5), 945-956 (2009-07-28)
Stinging nettle (Urtica dioica) is a herbaceous, dioecious perennial that is widely distributed around the world, reproduces both sexually and asexually, and is characterized by rapid growth. This work was aimed at evaluating the effects of plant maturity, shoot reproduction
Takeshi Kuroha et al.
Plant physiology and biochemistry : PPB, 43(5), 465-471 (2005-05-14)
To elucidate the physiological functions of the substances in xylem sap, we analyzed the biological activities of xylem sap from squash (Cucurbita maxima Duch.) root using tobacco BY-2 (Nicotiana tabacum L. cv. Bright Yellow 2) cell protoplasts. When BY-2 cell
Eloise Foo et al.
Plant physiology, 143(3), 1418-1428 (2007-02-06)
Increased-branching mutants of garden pea (Pisum sativum; ramosus [rms]) and Arabidopsis (Arabidopsis thaliana; more axillary branches) were used to investigate control of cytokinin export from roots in relation to shoot branching. In particular, we tested the hypothesis that regulation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service