Skip to Content
Merck
All Photos(4)

Key Documents

R8875

Sigma-Aldrich

Rotenone

≥95% (HPLC), powder, NADH-CoQ reductase inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22O6
CAS Number:
Molecular Weight:
394.42
Beilstein:
6773081
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Rotenone, ≥95%

Quality Level

Assay

≥95%

bp

210-220 °C/0.5 mmHg (lit.)

mp

159-164 °C (lit.)

SMILES string

COc1cc2OCC3Oc4c5C[C@@H](Oc5ccc4C(=O)C3c2cc1OC)C(C)=C

InChI

1S/C23H22O6/c1-11(2)16-8-14-15(28-16)6-5-12-22(24)21-13-7-18(25-3)19(26-4)9-17(13)27-10-20(21)29-23(12)14/h5-7,9,16,20-21H,1,8,10H2,2-4H3/t16-,20-,21+/m1/s1

InChI key

JUVIOZPCNVVQFO-HBGVWJBISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rotenone belongs to the retinoid family. It is naturally present in Leguminosa plants and is considered to be cytotoxic. It is commonly used as an insecticide and a pesticide.[1]

Application

Rotenone has been used:
  • to study its effect on cell proliferation[2]
  • to detect the production of cellular reactive oxygen species[3] in isolated rat forebrain mitochondria[4]
  • to measure mitochondrial enzymatic activity[5]

Biochem/physiol Actions

Inhibitor of mitochondrial electron transport.
Rotenone is an inhibitor of mitochondrial electron transport at nicotinamide adenine dinucleotide (NADH):ubiquinone oxidoreductase. It is readily absorbed through the exoskeletons of arthropods, but poorly absorbed cutaneously or from the gastrointestinal tract of mammals. Rotenone acts as a neurotoxic agent which can produce Parkinson-like condition to serve as an animal model for the study of etiology and interventions. The primary mechanism of action of rotenone is complex I inhibition, followed by the generation of oxidative stress and oxidative damage. Because rotenone is lipophilic, it readily penetrates the brain, where it attaches to and inhibits mitochondrial complex I of electron chain transport (ETC).

Preparation Note

This product is soluble in DMSO at 0.5 mg/ml. Other sources suggest it can be dissolved in DMSO up to 100 mM, however this has not been verified.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metformin decreases glucose oxidation and increases the dependency of prostate cancer cells on reductive glutamine metabolism
Fendt SM, et al.
Cancer Research (2013)
Parkinson's Disease and Movement Disorders, 81-81 (2007)
Loss of Sirt1 promotes prostatic intraepithelial neoplasia, reduces mitophagy, and delays PARK2 translocation to mitochondria
Di SG, et al.
The American Journal of Pathology, 185(1), 266-279 (2015)
Jin Bu et al.
Life sciences, 231, 116581-116581 (2019-06-21)
The aims of this study were to investigate the effect of colonic electrical stimulation (CES) on delayed colonic transit in Parkinson's disease (PD) model induced by rotenone and its possible mechanisms. Sprague-Dawley male rats were implanted with a pair of
Ca2+-induced oxidative stress in brain mitochondria treated with the respiratory chain inhibitor rotenone
Sousa SC, et al.
Febs Letters, 543(1-3), 179-183 (2003)

Questions

1–4 of 4 Questions  
  1. For how long can the product maintain stability in powder form at 4 °C or -80 °C?

    1 answer
    1. The recommended storage temperature for this material is ambient or room temperature. Studies have not been conducted to evaluate the stability or shelf life impact of the product when stored at other temperatures. In the original sealed container, the compound is quite stable at room temperature. Please see the link below to review additional information available in the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/366/380/r8875pis.pdf

      Helpful?

  2. Can aliquots of rotenone resuspended in DMSO be stored at -20C?

    1 answer
    1. The solution stability has not been determined. However, a similar product, 557368, notes that "following reconstitution, aliquot, flush with nitrogen, and store at -70°C. Stock solutions are stable for up to 1 month at -70°C".

      Helpful?

  3. What is the solubility in other compounds ?

    1 answer
    1. This product is tested for solubility at 50 mg/mL in chloroform. Rotenone is poorly soluble in water at approximately 0.25-0.50 uM. Other options include Ethanol and DMSO.
      Ethanol: 5 mg/mL (higher concentrations may be achieved with heat and sonication)
      DMSO: 50 mg/mL

      Helpful?

  4. What's the solubility in DMSO

    1 answer
    1. This product is soluble in DMSO at 0.5 mg/ml. Other sources suggest it can be dissolved in DMSO up to 100 mM, however this has not been verified.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service