Skip to Content
Merck
All Photos(1)

Key Documents

XAD16

Supelco

Amberlite XAD16N polymeric adsorbent

20-60 mesh

Synonym(s):

XAD16N

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52

Product Name

Amberlite XAD16N, 20-60 mesh

form

beads

technique(s)

LPLC: suitable

surface area

800 m2/g

matrix

styrene-divinylbenzene

matrix active group

polymer

particle size

20-60 mesh
560-710 μm

pore size

~0.55 mL/g pore volume
200 Å mean pore size

separation technique

reversed phase

SMILES string

[n]1(c(ccc1)CN2CCOCC2)C

InChI

1S/C10H16N2O/c1-11-4-2-3-10(11)9-12-5-7-13-8-6-12/h2-4H,5-9H2,1H3

InChI key

ZPPCOAPTXYJLTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.

Application

Hydrophobic compounds up to 40,000 MW; antibiotics; pharmaceutical manufacturing; surfactants; bitters; separation of large organic molecules (especially proteins). More efficient than XAD2.

Other Notes

Hydrophobic polyaromatic resin

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Don D Nguyen et al.
Nature microbiology, 2, 16197-16197 (2016-11-01)
Pseudomonads are cosmopolitan microorganisms able to produce a wide array of specialized metabolites. These molecules allow Pseudomonas to scavenge nutrients, sense population density and enhance or inhibit growth of competing microorganisms. However, these valuable metabolites are typically characterized one-molecule-one-microbe at
Shelja Tiwari et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 32(12), 1321-1325 (2016-12-13)
An online preconcentration system based on solid-phase extraction was developed for speciation and determinations of Cr in industrial water samples using flame atomic absorption spectrometry. A minicolumn packed with Amberlite XAD-16 loaded with salicylic acid was used to preconcentrate chromium
Katharina Rox et al.
Scientific reports, 7(1), 11798-11798 (2017-09-20)
In contrast to mild infections of Group A Streptococcus (GAS) invasive infections of GAS still pose a serious health hazard: GAS disseminates from sterile sites into the blood stream or deep tissues and causes sepsis or necrotizing fasciitis. In this
Shuqi Guo et al.
Microbial cell factories, 16(1), 203-203 (2017-11-17)
Xenocoumacin 1 (Xcn1) and Xenocoumacin 2 (Xcn2) are the main antimicrobial compounds produced by Xenorhabdus nematophila. Culture conditions, including pH, had remarkably distinct effects on the antimicrobial activity of X. nematophila. However, the regulatory mechanism of pH on the antimicrobial
Hirotoshi Sushida et al.
Journal of bioscience and bioengineering, 126(1), 23-29 (2018-02-18)
EnkT is a novel ATP-binding cassette (ABC) transporter responsible for secretion of four bacteriocins, enterocins NKR-5-3A, C, D, and Z (Ent53A, C, D, and Z), produced by Enterococcus faecium NKR-5-3. It is generally recognized that the secretion of a bacteriocin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service