Skip to Content
Merck
All Photos(4)

Key Documents

Z0164

Sigma-Aldrich

Zeatin

suitable for plant cell culture, BioReagent, powder

Synonym(s):

6-(4-Hydroxy-3-methylbut-2-enylamino)purine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O
CAS Number:
Molecular Weight:
219.24
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72
form:
powder
Assay:
≥98% (HPLC)

product line

BioReagent

Assay

≥98% (HPLC)

form

powder

technique(s)

cell culture | plant: suitable

color

off-white to yellow

application(s)

agriculture

storage temp.

−20°C

SMILES string

C\C(CO)=C/CNc1ncnc2[nH]cnc12

InChI

1S/C10H13N5O/c1-7(4-16)2-3-11-9-8-10(13-5-12-8)15-6-14-9/h2,5-6,16H,3-4H2,1H3,(H2,11,12,13,14,15)/b7-2+

InChI key

UZKQTCBAMSWPJD-FARCUNLSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Zeatin is a coconut milk-derived plant hormone and is a common plant cytokinin that is found in the developing or meristematic tissues and organs such as roots, shoot apex, immature organs, and others. Its structure includes the amino group of adenine being linked with the hydroxylated isoprene residue in the trans-position.

Biochem/physiol Actions

Zeatin promotes the growth of lateral buds and stimulates cell division (lateral dominance), callus initiation, and seed germination. Zeatin is used with Murashige and Skoog and other media. Zeatin promotes multiple shoot generation from internodes and high-frequency thick-spread roots.

Other Notes

Primarly trans isomer: balance cis isomer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanying Shen et al.
Horticulture research, 6, 51-51 (2019-05-10)
Growth monitoring indicated that the height of 'Kanshu' plants with 'Nantong-xiaofangshi' as an interstock was significantly shorter than that of 'Kanshu' plants with no interstock. A transcriptome analysis of the two graft combinations ('Kanshu'/Diospyros lotus and 'Kanshu'/'Nantong-xiaofangshi'/Diospyros lotus) was conducted
Xin-Wei Lee et al.
PloS one, 11(12), e0167958-e0167958 (2016-12-16)
Rafflesia is a biologically enigmatic species that is very rare in occurrence and possesses an extraordinary morphology. This parasitic plant produces a gigantic flower up to one metre in diameter with no leaves, stem or roots. However, little is known
Hanaé Roman et al.
Plant physiology, 172(1), 489-509 (2016-07-28)
Bud outgrowth is controlled by environmental and endogenous factors. Through the use of the photosynthesis inhibitor norflurazon and of masking experiments, evidence is given here that light acts mainly as a morphogenic signal in the triggering of bud outgrowth and
Ahmad Yamin Abdul Rahman et al.
BMC genomics, 14, 75-75 (2013-02-05)
Hevea brasiliensis, a member of the Euphorbiaceae family, is the major commercial source of natural rubber (NR). NR is a latex polymer with high elasticity, flexibility, and resilience that has played a critical role in the world economy since 1876.
Xiao-Ling Wang et al.
PloS one, 13(6), e0198878-e0198878 (2018-06-29)
The effect of plant population density (PPD) and root-induced leaf cytokinin on the compensatory growth of potted corn seedlings during post-drought rewatering was investigated. The study design comprised four treatments: (1) wetness with low PPD, (2) wetness with high PPD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service