Skip to Content
Merck
All Photos(3)

Documents

K3378

Sigma-Aldrich

Kinetin

suitable for plant cell culture, BioReagent, amorphous powder

Synonym(s):

6-Furfurylaminopurine, N6-Furfuryladenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9N5O
CAS Number:
Molecular Weight:
215.21
Beilstein:
21703
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

product line

BioReagent

Quality Level

Assay

≥98% (TLC and HPLC)

form

amorphous powder

technique(s)

cell culture | plant: suitable

color

off-white

mp

264-270 °C (dec.) (lit.)

application(s)

agriculture

SMILES string

C(Nc1ncnc2[nH]cnc12)c3ccco3

InChI

1S/C10H9N5O/c1-2-7(16-3-1)4-11-9-8-10(13-5-12-8)15-6-14-9/h1-3,5-6H,4H2,(H2,11,12,13,14,15)

InChI key

QANMHLXAZMSUEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kinetin is a cell division factor or cytokinin from DNA. It occurs naturally in various plant extracts such as those from coconuts, maize endosperm, banana and horsechestnut fruits, carrot and tobacco leaves, grapes, crown gall tumors, female gametophytes of Ginkgo and many other sources.

Application

Kinetin has been used as a supplement in modified MS medium for culturing various plant tissues. It has also been used to study its effect on thymus and immune function of aging rats.

Biochem/physiol Actions

Kinetin has an ability to induce cell division differentiation and growth. In addition, it also acts as an antioxidant and delays the aging process. Kinetin acts as a potential tool for studying effects of red light on plants.
Kinetin is an adenine-type cytokinin phytohormone that is used in plant culture media such as Murashige & Skoog media in conjunction with auxins such as indole-3-acetic acid (IAA), Indole-3-butyric acid (IBA), 1-napthaleneacetic acid (NAA) or others to induce the fomation of callus and to regenerate plant tissues from callus.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Similarity of some kinetin and red light effects.
Miller CO, et al.
Plant Physiology, 31(4), 318-318 (1956)
Jacek Łyczko et al.
Molecules (Basel, Switzerland), 26(9) (2021-05-01)
Rare carnivorous plants representing the genus Sarracenia are perceived as very interesting to scientists involved in various fields of botany, ethnobotany, entomology, phytochemistry and others. Such high interest is caused mainly by the unique capacity of Sarracenia spp. to attract
Effects of kinetin on thymus and immune function of aging rats
Li M, et al.
Protoplasma, 36(2), 356-362 (2016)
Stress induced acquisition of somatic embryogenesis in common bean Phaseolus vulgaris L.
Cabrera-Ponce JL, et al.
Protoplasma, 252(2), 559-570 (2015)
Kinetin and kinetin-like compounds
Modern Methods of Plant Analysis/Moderne Methoden der Pflanzenanalyse, 77(5), 194-202 (1963)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service