Skip to Content
Merck
All Photos(1)

Key Documents

P4333

Sigma-Aldrich

Penicillin-Streptomycin

liquid, non-animal origin, suitable for cell culture, BioReagent

Synonym(s):

Pen-Strep

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.76

Product Name

Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture

Quality Level

sterility

0.1 μm filtered

product line

BioReagent

form

liquid

concentration

100 ×

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes
protein synthesis | interferes

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC1(C)S[C@@H]2[C@H](NC(=O)Cc3ccccc3)C(=O)N2[C@H]1C(O)=O.CNC[C@H]4[C@H](O)[C@@H](O)[C@H](CCO)O[C@H]4O[C@H]5[C@@H](O[C@@H](C)C5(O)C=O)O[C@H]6[C@H](O)[C@@H](O)[C@H](NC(N)=N)[C@@H](O)[C@@H]6NC(N)=N

InChI

1S/C23H43N7O12.C16H18N2O4S/c1-7-23(38,6-32)18(42-19-8(5-28-2)12(33)13(34)9(40-19)3-4-31)20(39-7)41-17-11(30-22(26)27)14(35)10(29-21(24)25)15(36)16(17)37;1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9/h6-20,28,31,33-38H,3-5H2,1-2H3,(H4,24,25,29)(H4,26,27,30);3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22)/t7-,8-,9-,10+,11-,12-,13-,14+,15-,16+,17+,18-,19-,20-,23+;11-,12+,14-/m01/s1

InChI key

KMOOCZWLFBSQCW-XFOLRDBHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Penicillin Streptomycin solution, also known as Pen Strep, is the most commonly used antibiotic solution for the culture of mammalian cells. Penicillin Streptomycin solution is used to maintain sterile conditions during cell culture, and they show few adverse effects on the cultured cells.

Application

Penicillin-Streptomycin is used to supplement cell culture media to control bacterial contamination. It is recommended for use in cell culture applications at 10 mL/L.
Penicillin streptomycin solution has been used as a supplement in different types of media, including RGC (retinal ganglion cells) culture, α-MEM (minimum essential medium) growth medium, Dulbecco′s modified Eagle′s medium (DMEM)/nutrient mixture F-12 Ham, DMEM and DMEM/F-12 + GlutaMax.

Components

This product is formulated to contain 10,000 units penicillin and 10 mg streptomycin/mL solubilized in a proprietary citrate buffer.

Other Notes

20ml,100ml
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.Storage class (TRGS 510): Non Combustible Liquids

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ZhiMin Wang et al.
Transplantation, 103(3), 502-511 (2018-09-14)
Advances in immunosuppressive therapy have significantly improved short-term but not long-term survival of cardiac transplant recipients; this is largely due to severe cardiac allograft vasculopathy (CAV). Glucagon-like peptide-1 receptor (GLP-1R)-based therapy exerts physiological effects on the cardiovascular system in addition
J H Martínez-Liarte et al.
Pigment cell research, 8(2), 83-88 (1995-04-01)
Penicillin and streptomycin, the most widely used antibiotics in mammalian cell cultures, caused a moderate stimulation in dopa oxidase and tyrosine hydroxylase activities, but a slight inactivation in the dopachrome tautomerase activity of B16/F10 melanoma cells at the routine concentration
David Sabourin et al.
Journal of laboratory automation, 18(3), 212-228 (2012-09-28)
A microfluidic component library for building systems driving parallel or serial microfluidic-based assays is presented. The components are a miniaturized eight-channel peristaltic pump, an eight-channel valve, sample-to-waste liquid management, and interconnections. The library of components was tested by constructing various
184: Toxic effects of antibiotics routinely used in cell culture on primary cultures of dissociated mouse brain cells.
Ammon F
Pediatric Research, 10, 900-900 (1976)
Beinan Liu et al.
The Journal of biological chemistry, 286(6), 4248-4256 (2010-12-15)
We have previously shown that overexpressing subunits of the iron-binding protein ferritin can rescue the toxicity of the amyloid β (Aβ) peptide in our Drosophila model system. These data point to an important pathogenic role for iron in Alzheimer disease.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service