F7524
Fetal Bovine Serum
non-USA origin, sterile-filtered, suitable for cell culture
Synonym(s):
Fetal Calf sera, Fetal Calf serum, FBS, FCS, sera, serum
About This Item
Recommended Products
biological source
bovine fetus
Quality Level
sterility
sterile-filtered
product line
FBS Classic
composition
hemoglobin, ≤25 mg/dL
origin
non-USA origin
availability
not available in Canada, USA
technique(s)
cell culture | mammalian: suitable
impurities
≤10 EU/mL endotoxin
shipped in
dry ice
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Application
Biochem/physiol Actions
Features and Benefits
Analysis Note
- Endotoxin and hemoglobin tested
- Tested for the presence of bacteria, virus, and mycoplasma
- Triple filtered with 0.1 micron membrane under aseptic conditions
Legal Information
Not finding the right product?
Try our Product Selector Tool.
also commonly purchased with this product
recommended
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service