Skip to Content
Merck
All Photos(1)

Key Documents

T2501200

Tristearin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Glyceryl tristearate, 1,2,3-Trioctadecanoylglycerol, Glycerol tristearate, Tristearin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)16COOCH2]2CHOCO(CH2)16CH3
CAS Number:
Molecular Weight:
891.48
Beilstein:
1718456
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

tristearin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/C57H110O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h54H,4-53H2,1-3H3

InChI key

DCXXMTOCNZCJGO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tristearin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valentina Trotta et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 127, 250-259 (2018-02-28)
Lipid microparticles (LMs) uncoated or coated with chitosan and containing the neuroprotective polyphenol resveratrol were developed for its targeting to the brain via nasal administration. The lipid microparticles loaded with resveratrol (LMs-Res) were produced by melt emulsification, using stearic acid
Alam Zeb et al.
The Journal of pharmacy and pharmacology, 69(8), 955-966 (2017-04-20)
The aim of this study was to develop high payload itraconazole-incorporated lipid nanoparticles (HINP) with modulated release property using a binary mixture core of solid and liquid lipid for oral and parenteral administration. High payload itraconazole-incorporated lipid nanoparticles were prepared
Supandeep Singh Hallan et al.
Molecules (Basel, Switzerland), 25(6) (2020-03-27)
Ellagic acid (EA) is a potent antioxidant substance of natural origin characterized by poor biopharmaceutical properties and low solubility in water that limit its use. The aim of the present study was to develop lipid-based nanoparticle formulations able to encapsulate
Aidan McGlinchey et al.
Environment international, 143, 105935-105935 (2020-07-08)
In the last decade, increasing incidence of type 1 diabetes (T1D) stabilized in Finland, a phenomenon that coincides with tighter regulation of perfluoroalkyl substances (PFAS). Here, we quantified PFAS to examine their effects, during pregnancy, on lipid and immune-related markers
Nagavendra Kommineni et al.
Chemistry and physics of lipids, 224, 104707-104707 (2018-12-07)
Cabazitaxel as microtubule inhibitor and thymoquinone as HDAC inhibitor affects the important genes like p53, STAT3, Bax, BCL-2, p21 and down regulation of NF-κB are reported for potential activity against breast tumors. However, poor aqueous solubility and permeability hinders the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service