Skip to Content
Merck
All Photos(1)

Key Documents

T3924

Sigma-Aldrich

Trypsin-EDTA solution

1 ×, sterile; sterile-filtered, BioReagent, suitable for cell culture, 0.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red

Synonym(s):

Cell dissociation enzyme

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.78

biological source

Porcine

Quality Level

sterility

sterile; sterile-filtered

product line

BioReagent

form

solution

mol wt

23.4 kDa

concentration

1 ×

technique(s)

cell culture | mammalian: suitable
single cell analysis: suitable

impurities

Porcine parvovirus, none detected (9 CFR)

pH

7.0-9.0

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

The typical use for this product is in removing adherent cells from a culture surface. The concentration of trypsin necessary to dislodge cells from their substrate is dependent primarily on the cell type and the age of the culture.

Suitable for use in preparation of single cell suspension for sequencing.

Biochem/physiol Actions

Trypsin cleaves peptides on the C-terminal side of lysine and arginine residues. The rate of hydrolysis of this reaction is slowed if an acidic residue is on either side of the cleavage site and hydrolysis is stopped if a proline residue is on the carboxyl side of the cleavage site. The optimal pH for trypsin activity is 7-9. Trypsin can also act to cleave ester and amide linkages of synthetic derivatives of amino acids. EDTA is added to trypsin solutions as a chelating agent that neutralizes calcium and magnesium ions that obscure the peptide bonds on which trypsin acts. Removing these ions increases the enzymatic activity.

Serine protease inhibitors, including DFP, TLCK, APMSF, AEBSEF, and aprotinin, amongst others, will inhibit Trypsin.

Components

Trypsin consists of a single chain polypeptide of 223 amino acid residues, produced by the removal of the N-terminal hexapeptide from trypsinogen which is cleaved at the Lys - lle peptide bond. The sequence of amino acids is cross-linked by 6 disulfide bridges. This is the native form of trypsin, beta-trypsin. BETA-trypsin can be autolyzed, cleaving at the Lys - Ser residue, to produce alpha-trypsin. Trypsin is a member of the serine protease family.

Caution

This product is stored frozen between -10 and -40°C. Repeated cycles of freezing and thawing should be avoided.

Preparation Note

This product does contain phenol red. Due to shipment on dry ice, there could be significant carbon dioxide buildup in the package. This CO2 may enter the solution and lower the pH slightly, giving an orange rather than pinkish color. The orange solution will still be suitable for use, or the pH can be adjusted with sodium hydroxide. Incubating cells with too high a trypsin concentration for a long period can damage cell membranes and kill the cells. Solubilizing trypsin or diluting it from a concentrated solution should be done with a buffered salt solution containing no Ca2+ or Mg2+.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shiva Hamidian Jahromi et al.
Stem cells and development, 27(1), 44-54 (2017-11-11)
Tumor necrosis factor alpha (TNF-α) induced protein 6 is a major anti-inflammatory mediator released by activated mesenchymal stromal cells (MSCs). Neonatal MSCs are considered more metabolically active than cells derived from adult tissues, and potentially less heterogeneous. We hypothesized that
Ashraf Hassanpour et al.
Stem cell research & therapy, 9(1), 252-252 (2018-09-28)
The increasing number of patients with ovarian insufficiency due to autoimmune disorders, genetic predisposition, or iatrogenic effects of treatment such as cancer therapies necessitates an urgent measure to find a safe and transplantable alternative ovary. A bioengineered ovary is one
Zhiguo He et al.
Investigative ophthalmology & visual science, 57(15), 6639-6651 (2016-12-08)
Engineered corneal endothelial grafts able to provide numerous functional endothelial cells for the restoration of corneal transparency would be a worthwhile way of replacing donor tissue, which is extremely scarce. The grafts are simply constructed: a biocompatible thin and transparent
Ayşe Kübra Karaboğa Arslan et al.
Nutrients, 10(6) (2018-05-26)
The aim of this study is to investigate the potential inhibitory effect of α-chaconine and α-solanine on RL95-2 estrogen receptor (ER) positive human endometrial cancer cell line and to identify the effect of these glycoalkaloids on the Akt signaling and
Gloria Bonuccelli et al.
Oncotarget, 8(13), 20667-20678 (2017-02-23)
Here, we assembled a broad molecular "tool-kit" to interrogate the role of metabolic heterogeneity in the propagation of cancer stem-like cells (CSCs). First, we subjected MCF7 cells to "metabolic fractionation" by flow cytometry, using fluorescent mitochondrial probes to detect PCG1α

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service