Skip to Content
Merck
All Photos(1)

Key Documents

A6964

Sigma-Aldrich

Accutase® solution

sterile-filtered, suitable for cell culture

Synonym(s):

Cell detachment solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.75

sterility

sterile-filtered

Quality Level

form

liquid

concentration

400-600 units/mL

technique(s)

cell culture | mammalian: suitable
single cell analysis: suitable

impurities

≤1 EU/mL endotoxin

pH

6.8-7.8

shipped in

dry ice

storage temp.

−20°C

General description

Accutase® solution (in Dulbecco′s phosphate-buffered saline (DPBS) without Ca++ or Mg++) contains proteolytic and collagenolytic enzymes for cell detachment. It is useful for the routine detachment of cells from standard tissue culture plasticware and adhesion- coated plasticware. Accutase is preferred over trypsin for cell detachment as it helps to detach cells from culture vessels with less damage. Hence, Accutase is often used for the treatment of sensitive cells or for analyses of surface markers.

Application

Accutase® solution has been used for dissociation or digestion of nonadherent cells growing in an aggregated cluster, such as brain tumor cells, induced pluripotent stem cells (IPSCs) and glioblastoma cells into single cells.

Suitable for use in preparation of single cell suspension for sequencing.
Proven effective in detaching primary fibroblasts, endothelial cells, neurons, tumor cell lines, and insect cells. Performs exceptionally well in detaching cells for analysis of cell surface markers, virus growth assay, and flow cytometry as well as bioreactor scale-up. Does not contain mammalian or bacterial-derived products.

Features and Benefits

Ready-to-use liquid consisting of a proprietary mixture of proteolytic and collagenolytic enzymes for use in dissociation of cells from standard and adhesion-coated plasticware.

Formula variant

Prepared in Dulbecco′s PBS (0.2 g/L KCl, 0.2 g/L KH2PO4, 8 g/L NaCl, and 1.15 g/L Na2HPO4) containing 0.5 mM EDTA•4Na and 3 mg/L Phenol Red. Refer to Product Data Sheet on the website for product usage information.

Preparation Note

This product contains phenol red. The product is shipped on dry ice and there could be significant CO2 buildup in the package. This CO2 may enter the solution and lower the pH slightly giving a yellow-orange (around pH 6.8) color. The solution should still be good to use as is, or sodium hydroxide may be added to adjust the pH. A fully frozen bottle of Accutase will be yellow in color and change to an orange color as it defrosts. Once unpacked, the product can be refrozen or placed directly into the refrigerator for later use. Do not defrost the product in a warm water bath. Accutase is sensitive to temperatures above 37 °C and will be inactivated after 45 minutes at 37 °C. Defrosting overnight in the refrigerator or in a tub of cold tap water for 1-1.5 hours is recommended. Shake up the bottle of Accutase after defrosting to enable the ingredients to go into solution evenly. These products do not need to be warmed up before being used. Use promptly once removed from the refrigerator, and return to the refrigerator after use.

Legal Information

Accutase is a registered trademark of Innovative Cell Technologies, Inc.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent Zika virus isolates induce premature differentiation of neural progenitors in human brain organoids
Gabriel E, et al.
Cell Stem Cell, 20(3), 397-406 (2017)
Ruchi Bajpai et al.
Molecular reproduction and development, 75(5), 818-827 (2007-12-25)
Human embryonic stem cells (hESCs) hold great promise for cell-based therapies and drug screening applications. However, growing and processing large quantities of undifferentiated hESCs is a challenging task. Conventionally, hESCs are passaged as clusters, which can limit their growth efficiency
Autophagy pathways in glioblastoma
Jiang H, et al.
Methods in Enzymology, 453, 273-286 (2009)
Yanting Xue et al.
Stem cell research, 24, 151-154 (2017-10-17)
Human MSX1 gene is mapped to chromosome 4 and encodes a 303aa homeobox protein MSX1. MSX1 expression appears during early tooth development of vertebrate embryogenesis. Mutations in this protein are related to human tooth anomalie, cleft lip and palate and
DISC1 Regulates the Proliferation and Migration of Mouse Neural Stem/Progenitor Cells through Pax5, Sox2, Dll1 and Neurog2.
Qian Wu et al.
Frontiers in cellular neuroscience, 11, 261-261 (2017-09-14)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service