Skip to Content
Merck
All Photos(1)

Documents

CLS3516

Corning® Costar® TC-Treated Multiple Well Plates

size 6 wells, clear, polystyrene plate, flat bottom, case of 50 (individually wrapped), sterile, lid

Synonym(s):

cell culture plate, multi well plates, multiple well plates, multiwell plates, tissue culture plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
eCl@ss:
32190102
NACRES:
NA.75

material

clear
clear bottom
flat bottom
polystyrene plate
round clear wells

description

growth area 9.5 cm2

sterility

sterile

feature

lid
skirt

packaging

case of 50 (individually wrapped)

manufacturer/tradename

Corning 3516

size

6 wells

well diam.

34.8 mm

well volume

16.8 mL

well working volume

1.9-2.9 mL

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Rings on lid prevent cross-contamination. All plates have a uniform footprint and a raised bead to aid stacking. Alphanumerics provide well identification. Polystyrene unless stated otherwise.

Application

Corning® Costar® TC-Treated Multiple Well Plates have been used to:
  • cultivate SaOS-2cells
  • construct stable cell lines
  • seed and differentiate cells for real-time reverse transcription polymerase chain reaction (RT-PCR)

Features and Benefits

  • Flat bottoms for enhanced stability.
  • Specially treated to promote optimalcell attachment.
  • Ensured sterility through gammairradiation.
  • Free from pyrogens, ensuring a safe environmentfor cell culture.

Legal Information

Corning is a registered trademark of Corning, Inc.
Costar is a registered trademark of Corning, Inc.

recommended

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emad Tolba et al.
Journal of materials chemistry. B, 4(3), 376-386 (2016-01-21)
In human bone, amorphous calcium carbonate (ACC) is formed as a precursor of the crystalline carbonated apatite/hydroxyapatite (HA). Here we describe that the metastable ACC phase can be stabilized by inorganic polyphosphate (polyP) that is also used as a phosphate
Nhan Huynh et al.
Genome biology, 21(1), 279-279 (2020-11-19)
Advances in CRISPR technology have immensely improved our ability to manipulate nucleic acids, and the recent discovery of the RNA-targeting endonuclease Cas13 adds even further functionality. Here, we show that Cas13 works efficiently in Drosophila, both ex vivo and in
S Decembrini et al.
Scientific reports, 10(1), 10275-10275 (2020-06-26)
The development of improved methods to culture retinal organoids is relevant for the investigation of mechanisms of retinal development under pathophysiological conditions, for screening of neuroprotective compounds, and for providing a cellular source for clinical transplantation. We report a tissue-engineering
Chintha Lankatillake et al.
Plant methods, 17(1), 3-3 (2021-01-08)
Enzyme assays have widespread applications in drug discovery from plants to natural products. The appropriate use of blanks in enzyme assays is important for assay baseline-correction, and the correction of false signals associated with background matrix interferences. However, the blank-correction
Dakota B Ward et al.
International journal of molecular sciences, 20(17) (2019-08-24)
Contrast-induced acute kidney injury (CI-AKI) is the third most common cause of hospital associated kidney damage. Potential mechanisms of CI-AKI may involve diminished renal hemodynamics, inflammatory responses, and direct cytotoxicity. The hypothesis for this study is that diatrizoic acid (DA)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service