46944
Fluorescein isothiocyanate–dextran
average mol wt 4,000, (FITC:Glucose = 1:250)
Synonym(s):
FITC-Dextran 4, FITC–Dextran
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
mol wt
average mol wt 4,000
composition
(FITC:Glucose = 1:250)
extent of labeling
~0.004 mol/mol FITC glucose
solubility
H2O: 50 mg/mL, slightly hazy, orange
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Dextran labeled with fluorescein isothiocyanate for possible use in perfusion studies in animals.
FITC-dextran is used extensively in microcirculation and cell permeability research utilizing microfluorimetry. FITC-dextran has been used to study plant cell wall porosity and capillary permeability. Plasma proteins have been shown not to bind to FITC-dextran.
FITC-dextran is used in cardiovascular, microcirculation, perfusion, cell monolayer and cell membrane permeability research as fluorescent flux tracer compound that supports the measurement of processes such as blood flow, membrane damage, vascular drainage and renal elimination
Fluorescein isothiocyanate–dextran (FITC-Dextran 4) is use as a fluorescent probe to study cell permeability. FITC-Dextran 4 is used with to study permeability of epithelial cell such as intestinal and renal cells in vivo.
FITC-dextran is used in cardiovascular, microcirculation, perfusion, cell monolayer and cell membrane permeability research as fluorescent flux tracer compound that supports the measurement of processes such as blood flow, membrane damage, vascular drainage and renal elimination
Fluorescein isothiocyanate–dextran (FITC-Dextran 4) is use as a fluorescent probe to study cell permeability. FITC-Dextran 4 is used with to study permeability of epithelial cell such as intestinal and renal cells in vivo.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
related product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jhen-Wei Ruan et al.
Nature microbiology, 2, 16220-16220 (2016-11-29)
The gut microbiota plays profound roles in host metabolism and the inflammatory response associated with the development of obesity. Dusp6-deficient mice have been shown to be resistant to diet-induced obesity, but the mechanism behind this remains unclear. 16S ribosomal RNA
Development of a novel self-microemulsifying drug delivery system for reducing HIV protease inhibitor-induced intestinal epithelial barrier dysfunction.
Lei B, Zha W, Wang Y, Wen C, et al.
Molecular Pharmacology, 7, 844-853 (2010)
Adam J Moeser et al.
Research in veterinary science, 93(2), 975-982 (2012-01-31)
The objective of this study was to investigate intestinal function and morphology associated with peri-weaning failure to thrive syndrome (PFTS) in swine. Jejunum and distal ileum from control and pigs exhibiting PFTS was harvested at weaning, 4 and 11 days
Rebekah E Dumm et al.
Nature communications, 10(1), 779-779 (2019-02-17)
Influenza B virus (IBV) is an acute, respiratory RNA virus that has been assumed to induce the eventual death of all infected cells. We and others have shown however, that infection with apparently cytopathic viruses does not necessarily lead to
Akira Ikari et al.
Journal of cellular physiology, 226(6), 1471-1478 (2010-08-19)
Migration of renal epithelial cells increases after renal tubular damage, but its mechanism has not been clarified in detail. Hyperosmotic stress increased a cellular injury concomitant with a decrease in mRNA and protein expression of claudin-2 in renal tubular epithelial
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service