Skip to Content
Merck
All Photos(2)

Documents

P7626

Sigma-Aldrich

Phenylmethanesulfonyl fluoride

powder, ≥98.5% (GC)

Synonym(s):

α-Toluenesulfonyl fluoride, Benzylsulfonyl fluoride, PMSF, Phenylmethylsulfonyl fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7FO2S
CAS Number:
Molecular Weight:
174.19
Beilstein:
2088311
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

product name

Phenylmethanesulfonyl fluoride, ≥98.5% (GC)

biological source

synthetic

Assay

≥98.5% (GC)

form

powder

mp

91-94 °C

solubility

dry solvents (ethanol, methanol, and 2-propanol): 200 mM (Stock solution are stable for months at 4°C.)
H2O: unstable

SMILES string

FS(=O)(=O)Cc1ccccc1

InChI

1S/C7H7FO2S/c8-11(9,10)6-7-4-2-1-3-5-7/h1-5H,6H2

InChI key

YBYRMVIVWMBXKQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenylmethanesulfonyl fluoride (PMSF) is a widely used serine protease inhibitor, effective against such enzymes as chymotrypsin, thrombin, and trypsin. PMSF acts as an inhibitor via sulfonation of the hydroxyl residues of serine residues at the reactive sites of serine proteases.

Application

PMSF is often used in lysis buffer to assist in preserving proteins of interest during protein isolation and sample preparation, by inhibiting proteases that would otherwise degrade proteins after cell or tissue lysis. PMSF is unstable in aqueous media. Thus stock solutions of PMSF are generally prepared in anhydrous organic solvents (e.g. 100% ethanol, or anhydrous isopropanol) prior to use in aqueous media.

Noted general features and benefits of PMSF include the following:
  • Inhibits serine proteases such as trypsin and chymotrypsin
  • Also inhibits cysteine proteases (reversible by reduced thiols) and mammalian acetylcholinesterase
  • Not as effective or as toxic as DFP
  • Effective concentration 0.1-1 mM
  • Half-life = 1 hr. at pH 7.5
Phenylmethanesulfonyl fluoride has been used in following applications:
  • cell fractionation.
  • used as a supplement in nuclear protein extraction.
  • inhibitor of cholesterol esterase (CE) and pseudocholinesterase (PCE).
  • used for the collection of blood prior to centrifugation to quantify plasma ANP levels.

Biochem/physiol Actions

Administration of PMSF produces analgesia unrelated to its anticholinesterase effect, and prolongs the analagesic effect of centrally administered β-endorphin.

related product

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Salivary esterase activity and its association with the biodegradation of dental composites.
Finer Y and Santerre JP.
Journal of Dental Research, 83(1), 22-26 (2004)
Estrogen receptors activate atrial natriuretic peptide in the rat heart.
Jankowski M, et al.
Proceedings of the National Academy of Sciences of the USA, 98(20), 11765-11770 (2001)
C Pinsky et al.
Life sciences, 31(12-13), 1193-1196 (1982-09-20)
Intraperitoneal (IP) injection of the serine proteinase inhibitor phenylmethylsulfonyl fluoride (PMSF) produced dose-dependent analgesia in Sprague-Dawley rats. AD50 was 2.9 +/- 1.4 (S.E.) mg kg-1, the analgesia was antagonized by naloxone but unaffected by atropine. PMSF significantly enhanced the analgesic
Lise Boussemart et al.
Nature, 513(7516), 105-109 (2014-08-01)
In BRAF(V600)-mutant tumours, most mechanisms of resistance to drugs that target the BRAF and/or MEK kinases rely on reactivation of the RAS-RAF-MEK-ERK mitogen-activated protein kinase (MAPK) signal transduction pathway, on activation of the alternative, PI(3)K-AKT-mTOR, pathway (which is ERK independent)
Löms Ziegler-Heitbrock et al.
Journal of immunology (Baltimore, Md. : 1950), 171(1), 285-290 (2003-06-21)
The anti-inflammatory cytokine IL-10 can be induced by type I IFNs, but the molecular mechanisms involved have remained elusive. With in silico analysis of the human IL-10 promoter we identified a module consisting of an IFN regulatory factor 1 (IRF-1)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service