Skip to Content
Merck
All Photos(4)

Documents

B6506

Sigma-Aldrich

Benzamidine hydrochloride hydrate

≥99%

Synonym(s):

Amidinobenzene hydrochloride, Benzamidinium chloride, Benzenecarboximidamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(=NH)NH2 · HCl · xH2O
CAS Number:
Molecular Weight:
156.61 (anhydrous basis)
Beilstein:
3594959
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥99%

form

powder

mp

86-88 °C (lit.)

solubility

water: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

Cl[H].[H]O[H].NC(=N)c1ccccc1

InChI

1S/C7H8N2.ClH.H2O/c8-7(9)6-4-2-1-3-5-6;;/h1-5H,(H3,8,9);1H;1H2

InChI key

BQVJCMMLDJPBFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Benzamidine hydrochloride hydrate has been used:
  • in western blot analysis
  • in immunoprecipitation
  • in the purification of yeast dynein
  • as a protease inhibitor in the preparation of brush-border membrane vesicles
  • for the processing of human synovial fluid, blood and eye sleep
  • to prevent non-specific degradation of cartilages

Biochem/physiol Actions

Benzamidine HCl is a reversible inhibitor of trypsin, trypsin-like enzymes, and serine proteases. A concentration of approximately 1 mM is used for general protease inhibition. To inhibit proteases from yeast, a range of 0.5 to 4.0 mM is used and it is for the most part interchangeable with pepstatin A.

In addition to being a strong competitive inhibitor of trypsin, benzamidine HCl has been also shown to be a strong competitive inhibitor of thrombin and plasmin. It was also found to be as effective as aprotinin in the prevention of glucagon degradation in human plasma.
Peptidase inhibitor.

Preparation Note

Benzamidine HCl is soluble in water and alcohol. Solubility testing in water at a concentration of 50 mg/ml yields a clear solution with heating. Benzamidine HCl is sensitive to oxidation. It is recommended to prepare solutions fresh each time in degassed water prior to use. However, frozen aliquots stored under inert gas, to exclude air, may be stable for a short time. Insufficient information is available to assess the shelf-life of a frozen solution.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aneuploidy causes premature differentiation of neural and intestinal stem cells
Gogendeau D, et al.
Nature Communications, 6(51), 8894-8894 (2015)
Sialidase unmasks mucin domain epitopes of lubricin
Solka KA, et al.
The Journal of Histochemistry and Cytochemistry, 64(11), 647-668 (2016)
Contrast-enhanced CT imaging as a non-destructive tool for ex vivo examination of the biochemical content and structure of the human meniscus
Oh DJ, et al.
Journal of Orthopaedic Research : Official Publication of the Orthopaedic Research Society, 35(5), 1018-1028 null
Contrast-enhanced CT using a cationic contrast agent enables non-destructive assessment of the biochemical and biomechanical properties of mouse tibial plateau cartilage
Lakin BA, et al.
Journal of Orthopaedic Research : Official Publication of the Orthopaedic Research Society, 34(7), 1130-1138 (2016)
Combining Structure-Function and Single-Molecule Studies on Cytoplasmic Dynein
Single Molecule Analysis, 53-89 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service