Skip to Content
Merck
All Photos(2)

Key Documents

A8456

Sigma-Aldrich

4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride

≥97.0% (HPLC)

Synonym(s):

AEBSF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10FNO2S · HCl
CAS Number:
Molecular Weight:
239.69
Beilstein:
7604627
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥97.0% (HPLC)

form

powder

mp

183-191  °C

solubility

H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

FS(C1=CC=C(CCN)C=C1)(=O)=O.[H]Cl

InChI

1S/C8H10FNO2S.ClH/c9-13(11,12)8-3-1-7(2-4-8)5-6-10;/h1-4H,5-6,10H2;1H

InChI key

WRDABNWSWOHGMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

stable for up to six months if stored refrigerated at a pH of less than 7. If a pH of greater than 7 is required, pH adjustment should be made just prior to use.

Application

4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride (AEBSF) has been used:
  • as a protease inhibitor in plasma samples to prevent acylated ghrelin (AG) degradation
  • as a snake venom serine proteinase (SVSP) inhibitor in SVSP inhibition or AEBSF assay to test the involvement of SVSPs on the fibrinogen-clotting activity
  • as a component in NP-40 lysis buffer to resuspend cell pellets for preparing cell lysates

Recommended concentrations for use are 0.1-1 mM.

Biochem/physiol Actions

4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride (AEBSF) is an irreversible serine protease inhibitor. Inhibition constants are similar to those of phenylmethanesulfonyl fluoride (PMSF) and diisopropylfluorophosphate (DFP). AEBSF has been shown to inhibit trypsin, chymotrypsin, plasmin kallikrein, and thrombin. As an alternative to PMSF and DFP, AEBSF offers lower toxicity, improved solubility in water, and improved stability in aqueous solutions. AEBSF can be used in cell culture in concentrations of up to 0.25 mM.
AEBSF can inhibit acetylcholinesterase (AChE).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilario Mennella et al.
The Journal of nutrition, 145(9), 2169-2175 (2015-07-17)
Food palatability increases food intake and may lead to overeating. The mechanisms behind this observation are still largely unknown. The aims of this study were the following: 1) to elucidate the plasma responses of endocannabinoids, N-acylethanolamines, and gastrointestinal peptides to
Ignacio Pedrós et al.
Biochimica et biophysica acta, 1842(9), 1556-1566 (2014-06-03)
The present study had focused on the behavioral phenotype and gene expression profile of molecules related to insulin receptor signaling in the hippocampus of 3 and 6 month-old APPswe/PS1dE9 (APP/PS1) transgenic mouse model of Alzheimer's disease (AD). Elevated levels of
Xue Zhang et al.
Frontiers in plant science, 11, 277-277 (2020-03-29)
Post-translational covalent modifications of histones play important roles in modulating chromatin structure and are involved in the control of multiple developmental processes in plants. Here we provide insight into the contribution of the histone lysine methyltransferase SET DOMAIN GROUP 8
Amy M Weeks et al.
Current protocols in chemical biology, 12(1), e79-e79 (2020-02-20)
Subtiligase is a powerful enzymatic tool for N-terminal modification of proteins and peptides. In a typical subtiligase-catalyzed N-terminal modification reaction, a peptide ester donor substrate is ligated onto the unblocked N terminus of a protein, resulting in the exchange of
Niina Aaltonen et al.
Biological procedures online, 22, 6-6 (2020-03-20)
Serine hydrolases (SHs) are a functionally diverse family of enzymes playing pivotal roles in health and disease and have emerged as important therapeutic targets in many clinical conditions. Activity-based protein profiling (ABPP) using fluorophosphonate (FP) probes has been a powerful

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service