Skip to Content
Merck
All Photos(1)

Key Documents

52332

Sigma-Aldrich

Phenylmethylsulfonyl Fluoride

Phenylmethylsulfonyl Fluoride, CAS 329-98-6, is an irreversible inhibitor of serine proteases. It causes sulfonylation of the active-site serine residues.

Synonym(s):

Phenylmethylsulfonyl Fluoride, Benzylsulfonyl Fluoride, PMSF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7FO2S
CAS Number:
Molecular Weight:
174.19
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

description

RTECS - XT8040000

Assay

≥99% (GC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white to off-white

solubility

ethanol: soluble
isopropanol: soluble
methanol: soluble

shipped in

ambient

storage temp.

10-30°C

SMILES string

F[S](=O)(=O)Cc1ccccc1

InChI

1S/C7H7FO2S/c8-11(9,10)6-7-4-2-1-3-5-7/h1-5H,6H2

InChI key

YBYRMVIVWMBXKQ-UHFFFAOYSA-N

General description

Irreversible inhibitor of serine proteases. Its mechanism of action is analogous to that of diisopropylfluorophosphate. PMSF causes sulfonylation of the active-site serine residues. Also reported to inhibit internucleosomal DNA fragmentation in immature thymocytes. For a related, more stable inhibitor, see AEBSF (Cat. No. 101500).

Biochem/physiol Actions

Cell permeable: no
Primary Target
serine proteases
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Corrosive (L)

Reconstitution

Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 9 months at 4°C.
t½ = 1 hour at pH 7.5. Effective concentration: 50 μM.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Seitz, R., et al. 1993. Int. J. Cancer53, 514.
Weaver, V.M., et al. 1993. Biochem. Cell. Biol.71, 488.
Bourgain, R.H., et al. 1992. Adv. Exp. Med. Biol.316, 427.
Chang, C.T., et al. 1992. Biochem. Int.28, 707.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrizio Clarelli et al.
PLoS computational biology, 16(8), e1008106-e1008106 (2020-08-17)
Antibiotic resistance is rising and we urgently need to gain a better quantitative understanding of how antibiotics act, which in turn would also speed up the development of new antibiotics. Here, we describe a computational model (COMBAT-COmputational Model of Bacterial
Xuejing Ma et al.
Biophysics reports, 9(1), 3-14 (2023-07-10)
Adipose triacylglycerol lipase (ATGL) is a dynamic lipid droplet-associated protein involved in cellular lipolysis, which is conserved from bacteria to humans. Recent methods that measure the enzymatic activity of ATGL in vitro are established using lipid emulsions. However, the lipid
Proximity Protein Labeling In Dictyostelium With Engineered Ascorbic Acid Peroxidase 2.
Takashima, et al.
Journal of biological methods, 10, e99010002-e99010002 (2023)
Siqi Ming et al.
Frontiers in immunology, 12, 736269-736269 (2021-10-01)
Gastric CD4+T cells contribute to Helicobacter pylori (H. pylori)-induced gastritis by amplifying mucosal inflammation and exacerbating mucosal injuries. However, the pathogenic CD4+ T cell subset involved in gastritis and the potential regulators are still unclear. Here we identified an IL-21-producing
Ming Chang et al.
Bio-protocol, 7(8), e2227-e2227 (2017-04-20)
Arabidopsis pollen is an excellent system for answering important biological questions about the establishment and maintenance of cellular polarity and polar cell growth, because these processes are amenable to the genetic and genomic approaches that are readily available in Arabidopsis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service