Skip to Content
MilliporeSigma

Skip To

CO-RO

Roche

cOmplete Protease Inhibitor Cocktail

Tablets provided in glass vials

Synonym(s):

protease inhibitor tablet

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 tabs
Check Cart for Availability
$562.00
3 x 20 tabs
Check Cart for Availability
$1,540.00
$1,355.20

About This Item

UNSPSC Code:
12352204
NACRES:
NA.54
Form:
tablet
Solubility:
aqueous buffer: soluble
Storage temp.:
2-8°C

$562.00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

tablet

packaging

vial of 20 tablets (11697498001), vial of 3 × 20 tablets (11836145001)

manufacturer/tradename

Roche

optimum pH

<7.5

solubility

aqueous buffer: soluble

absorption

0.08 at 280 nm

storage temp.

2-8°C

General description

cOmplete Protease Inhibitor Tablets inhibit a broad spectrum of serine, cysteine, and metalloproteases, as well as calpains. Due to the optimized composition of the tablets, they show excellent inhibition effects, and are well suited for the protection of proteins isolated from animal tissues, plants, yeast, and bacteria. cOmplete tablets contain EDTA and both irreversible and reversible protease inhibitors.

Application

cOmplete is used for the inhibition of serine, cysteine, and metalloproteases in bacterial, mammalian, yeast, and plant cell extracts.[1][2][3][4][5]
cOmplete contains both reversible and irreversible protease inhibitors. Therefore, we recommend the addition of cOmplete to all stock buffers and solutions normally protected with protease inhibitors, and not  just during the initial purification steps.
cOmplete contains EDTA (1 tablet yields a  1 mM EDTA solution in 50 ml). Therefore, the extraction buffer should not contain divalent cations like Ca2+ , Mg2+, or Mn2+, otherwise the inhibition of the metalloproteases might be incomplete. If the protein of interest will be purified by IMAC (Immobilized Metal Affinity Chromatography), for example, Poly-His-tagged recombinant proteins, EDTA has to be eliminated (e.g., by dialysis) prior to the chromatography.
Alternatively, the product cOmplete EDTA-free can be used. These tablets are identical to cOmplete, with the exception that no EDTA or other chelating agent is  present.
cOmplete Tablets inhibit proteolytic activity in large extraction volumes (up to 50 ml). If you are working with smaller volumes (up to 10 ml), we recommend to use cOmplete, Mini. The composition of the cOmplete, Mini Tablet is identical to the normal cOmplete Tablet.
cOmplete Protease Inhibitor Cocktail has been used as a component of
  • lysis buffer for the homogenisation of rat cavernosal smooth muscle (CSM) for obtaining membranous and cytosolic fractions
  • cold buffer A for preparing cytosolic and nuclear extracts
  • lysis buffer for homogenisation of tissues for determining protein expression by western blot
  • phosphate buffered saline (PBS) for the lysis of rat retinae for enzyme linked immunosorbent assay (ELISA)

Biochem/physiol Actions

Mixture of several protease inhibitors with broad inhibitory specificity. For the inhibition of serine, cysteine, and metalloproteases in bacterial, mammalian, yeast, and plant cell extracts.

Features and Benefits

Use cOmplete Protease Inhibitor Tablets to protect your proteins from a wide range of proteases. In just minutes, uninhibited proteolytic activity can degrade the protein you have spent days isolating. Each of these convenient water-soluble tablets contains a blend of protease inhibitors that inhibits proteolytic activity from most cell types, including animals, plants, yeast, and bacteria. No weighing or measuring is necessary, ensuring consistent results.

  • Easy-to-use: Simply drop a quick-dissolving tablet into your buffer.
  • Complete protection: Instantly protect your proteins against a broad range of proteases.
  • Flexible: Protect proteins in extracts from almost any tissue or cell, including animals, plants, yeast, bacteria, or fungi.
  • Safe: Choose non-toxic inhibitors that pose no risk to you or those around you.
  • Suitable for IMAC purification: Purify poly-histidine tagged proteins with cOmplete His-Tag Purification Resin.

Physical form

Tablets in glass vials

Preparation Note

Working concentration: 1 tablet per 50 ml extraction solution
Working solution: Preparation of Working Solution:

One cOmplete Protease Inhibitor Tablet is sufficient for the inhibition of the proteolytic activity in 50 ml extraction solution. If very high proteolytic activity is present, one tablet should be used for 25 ml extraction buffer. Tablets can be added directly to the extraction medium. Alternatively, a stock solution (25 X conc.) can be prepared.

Preparation of Stock solution (25 X conc.):

Dissolve one cOmplete Protease Inhibitor tablet in 2 ml dist. water or in 2 ml 100 mM phosphate buffer, pH 7.0. The stock solution can also be prepared in 10 mM Tris buffer, pH 7.2 to 7.5.
Storage conditions (working solution): The stock solution can be stored at 2 to 8 °C for 1 to 2 weeks, or at least 12 weeks at -15 to -25 °C.
Soluble in aqueous buffers, or add directly to extraction media. Alternatively, prepare 25x stock solutions in 2 ml water or 100 mM phosphate buffer, pH 7.0. Stock solution is stable for 1 to 2 weeks at 2 to 8 °C or at least 12 weeks at -15 to -25 °C; can also be used in thiol-containing solutions at 15 to 25 °C.

Analysis Note

Performance-tested with pancreas extract.

Other Notes

Inhibition efficiency is quickly and sensitively tested with the Universal Protease Substrate. For life science research only. Not for use in diagnostic procedures.
Proprietary mixture of several protease inhibitors with broad inhibitory specificity.

Legal Information

cOmplete is a trademark of Roche

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
046931160010469312400104693132001
form

tablet

form

tablet

form

tablet

form

tablet

manufacturer/tradename

Roche

manufacturer/tradename

Roche

manufacturer/tradename

Roche

manufacturer/tradename

Roche

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

packaging

vial of 20 tablets (11697498001), vial of 3 × 20 tablets (11836145001)

packaging

pkg of 20 tablets

packaging

pack of 30 tablets (individually packed in foil blister packs)

packaging

pkg of 20 tablets (individually packed in foil blister packs)

solubility

aqueous buffer: soluble

solubility

water: soluble

solubility

aqueous buffer: soluble, water: soluble

solubility

aqueous buffer: soluble, water: soluble

absorption

0.08 at 280 nm

absorption

0.08 at 280 nm

absorption

-

absorption

0.08 at 280 nm


Still not finding the right product?


signalword

Danger

wgk

WGK 2

flash_point_f

does not flash

flash_point_c

does not flash

pictograms

CorrosionHealth hazard

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT RE 2

Storage Class

8A - Combustible corrosive hazardous materials



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library