Skip to Content
Merck
All Photos(1)

Key Documents

78830

Sigma-Aldrich

Phenylmethanesulfonyl fluoride

≥99.0% (T)

Synonym(s):

α-Toluenesulfonyl fluoride, Benzylsulfonyl fluoride, PMSF, Phenylmethylsulfonyl fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7FO2S
CAS Number:
Molecular Weight:
174.19
Beilstein:
2088311
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Assay

≥99.0% (T)

form

powder or crystals

impurities

≤0.1% water

mp

92-95 °C

solubility

anhydrous isopropanol: soluble 35 mg/mL, clear to faintly hazy, colorless to faintly yellow (Heating required)

storage temp.

room temp

SMILES string

FS(=O)(=O)Cc1ccccc1

InChI

1S/C7H7FO2S/c8-11(9,10)6-7-4-2-1-3-5-7/h1-5H,6H2

InChI key

YBYRMVIVWMBXKQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phenylmethanesulfonyl fluoride has been used as a component in radioimmunoprecipitation buffer for western blot analysis. It has also been used as a component in the RAW264.7 cell lysis buffer for western blotting.

Biochem/physiol Actions

Phenylmethanesulfonyl fluoride has the ability to enhance the stability of plasma lipidome in lipidomic and metabolomic analysis. This serine protease inhibitor reduces the naloxone-precipitated withdrawal jumping behavior in morphine-dependent mice.

Other Notes

Specific trypsin and chymotrypsin inhibitor. Less toxic substitute for diisopropylfluorophosphate. Potentiates the effects of ischemia on brain mitochondria through its inhibition of phospholipase C.
Specific trypsin and chymotrypsin inhibitor; Less toxic substitute for diisopropylfluorophosphate. Potentiates the effects of ischemia on brain mitochondria through its inhibition of phospholipase C.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Q S Wang et al.
Acta pharmacologica Sinica, 22(3), 249-252 (2001-12-18)
To study the effect of phenylmethylsulfonyl fluoride (PMSF), a phospholipase C inhibitor, on ischemic injury to brain mitochondria in rats. The phospholipid level, membrane fluidity, and respiratory control ratio of brain mitochondria were measured. The effect of phenylmethylsulfonyl fluoride was
Phenylmethanesulfonyl fluoride pretreatment stabilizes plasma lipidome in lipidomic and metabolomic analysis
Wang X, et al.
Analytica Chimica Acta, 893, 77-83 (2015)
Cellular bioenergetics changes in magnocellular neurons may affect copeptin expression in the late phase of sepsis
Oliveira-Pelegrin G R, et al.
Journal of Neuroimmunology, 267(1-2), 28-34 (2014)
beta-catenin dosage is a critical determinant of tracheal basal cell fate determination
Brechbuhl H M, et al.
The American Journal of Pathology, 179(1), 367-379 (2011)
Quercetin protects RAW264. 7 macrophages from glucosamine-induced apoptosis and lipid accumulation via the endoplasmic reticulum stress pathway
Cai X, et al.
Molecular Medicine Reports, 12(5), 7545-7553 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service