Skip to Content
Merck
All Photos(1)

Documents

P2192

Sigma-Aldrich

10X PCR Buffer

Optimized for routine PCR with MgCl2 included

Synonym(s):

PCR Buffer Solution, PCR Reaction Buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106306
NACRES:
NA.52

Quality Level

form

liquid

packaging

vial of 1.5 mL

color

colorless

application(s)

agriculture

foreign activity

DNase, RNase, none detected

storage temp.

−20°C

Application

10X PCR Buffer has been used as a component of the PCR mixture:
  • for the amplification of viral DNA
  • to amplify genomic DNA to study the methylenetetrahydrofolate reductase (MTHFR) gene mutations in attention deficit hyperactivity disorder (ADHD) individuals
  • to amplify 16S-23S rRNA gene internal transcribed spacer (ITS) from Bacillus sp.

Features and Benefits

  • Standalone buffer
  • Compatible with JumpStart Taq DNA Polymerase (D9307), Taq DNA Polymerase from Thermus aquaticus (D1806), and REDTaq® Genomic DNA Polymerase (D8312)

Components

Composition of the 10× buffer: 100 mM Tris-HCl, pH 8.3 at 25°C; 500 mM KCl; 15 mM MgCl2; 0.01% gelatin

Legal Information

JumpStart is a trademark of Sigma-Aldrich Co. LLC
REDTaq is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phylogenetic Analysis of Baculovirus Isolates from Diseased Insects in Southern Vietnam
Thao N T P, et al.
Open Journal of Genetics, 2014 (2014)
Maike Stam et al.
Genetics, 162(2), 917-930 (2002-10-26)
Paramutation is an interaction between alleles that leads to a heritable change in the expression of one allele. In B'/B-I plants, B-I (high transcription) always changes to B' (low transcription). The new B' allele retains the low expression state in
Ákos Boros et al.
PloS one, 6(12), e29145-e29145 (2011-12-24)
A novel positive-sense, single-stranded RNA (+ssRNA) virus (Halastavi árva RNA virus, HalV; JN000306) with di-cistronic genome organization was serendipitously identified in intestinal contents of freshwater carps (Cyprinus carpio) fished by line-fishing from fishpond "Lőrinte halastó" located in Veszprém County, Hungary.
G J Breedveld et al.
Neurology, 59(4), 579-584 (2002-08-28)
Benign hereditary chorea (BHC) is an autosomal dominant disorder that can be distinguished from Huntington disease by its early onset, stable or only slightly progressive course, and absence of mental deterioration. The variation in clinical features is such that its
Cem Gokcen et al.
International journal of medical sciences, 8(7), 523-528 (2011-09-08)
The purpose of this study was to evaluate the relationship between 5,10- methylenetetrahydrofolate reductase (MTHFR) polymorphisms and Attention Deficit Hyperactivity Disorder (ADHD) in a sample of Turkish children. MTHFR gene polymorphisms were assessed in 40 patients with ADHD and 30

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service