D4309
REDTaq® DNA Polymerase
Taq for routine PCR with inert dye, 10X buffer included
Synonym(s):
DNA polymerase with loading dye, Taq DNA polymerase
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Recommended Products
form
liquid
usage
sufficient for 1000 reactions
sufficient for 250 reactions
sufficient for 50 reactions
feature
dNTPs included: no
hotstart: no
concentration
1 unit/μL
technique(s)
PCR: suitable
color
red
input
purified DNA
suitability
suitable for PCR
application(s)
agriculture
shipped in
wet ice
storage temp.
−20°C
Related Categories
General description
REDTaq® DNA Polymerase is a unique blend of Sigma′s quality Taq DNA Polymerase combined with an inert red dye. This dye enables quick visual confirmation of enzyme addition and reaction mixing. An aliquot of the samples (5-10 μl) can then be loaded directly onto an agarose gel for electrophoresis following PCR. The red dye migrates slightly faster than bromophenol blue at about the same rate as a 125 base pair fragment in a 1% agarose gel. Since no additional loading buffers are added to the reaction following PCR, reamplification is possible.
The red dye has no effect on automated or manual sequencing, restriction digestions or other downstream applications. However, if removing the dye is desired, this can easily be accomplished using any standard purification method.
The red dye has no effect on automated or manual sequencing, restriction digestions or other downstream applications. However, if removing the dye is desired, this can easily be accomplished using any standard purification method.
Application
For routine PCR amplifications REDTaq® DNA Polymerase has been used in polymerase chain reaction (PCR) and reverse transcription-polymerase chain reaction (RT-PCR) analysis.
Features and Benefits
- Same great performance as Taq DNA Polymerase in a more convenient format for high throughput applications.
- Visual confirmation that not only has the enzyme been added, but that proper mixing has occurred.
- No additional loading dyes are necessary. An aliquot can be taken directly from the reaction and loaded onto an agarose gel for electrophoresis.
Packaging
Provided with 10X reaction buffer containing MgCl2
Unit Definition
One unit incorporates 10 nmol of total dNTPs into acid-precipitable DNA in 30 min at 74 °C.
Legal Information
Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: US 8,404,464 and US 7,972,828. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims.
REDTaq is a registered trademark of Merck KGaA, Darmstadt, Germany
recommended
related product
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class Code
10 - Combustible liquids
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Mutation or polymorphism of the CD20 gene is not associated with the response to R-CHOP in diffuse large B cell lymphoma patients
Sar A, et al.
Leukemia Research, 33(6), 792-797 (2009)
Mapping networks of protein-mediated physical interactions between chromitin elements.
Tiwari, V.K., and Baylin, S.B.
Current Protocols in Molecular Biology, 21-21 (2010)
Proteomics-based approach to elucidate the mechanism of antitumor effect of curcumin in cervical cancer
Madden K, et al.
Prostaglandins, Leukotrienes, and Essential Fatty Acids, 80(1), 9-18 (2009)
Ovidiu Paun et al.
Methods in molecular biology (Clifton, N.J.), 862, 75-87 (2012-03-16)
Amplified fragment length polymorphism (AFLP) is a PCR-based technique that uses selective amplification of a subset of digested DNA fragments to generate and compare unique fingerprints for genomes of interest. The power of this method relies mainly in that it
Kaleigh A Suhs et al.
mBio, 2(3), e00094-e00011 (2011-06-02)
Toll-like receptor 4 is thought to have a primary role in host defense against Escherichia coli bladder colonization, based on mouse models of urinary tract infection using C3H/HeJ female mice. This strain carries a point mutation in the Tlr4 gene
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service