Skip to Content
Merck
All Photos(3)

Key Documents

M8266

Sigma-Aldrich

Magnesium chloride

anhydrous, ≥98%

Synonym(s):

Magnogene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCl2
CAS Number:
Molecular Weight:
95.21
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥98%
grade:
anhydrous
form:
powder
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

Assay

≥98%

form

powder

impurities

≤0.5% Insolubles

bp

1412 °C/1 atm (lit.)

mp

714 °C (lit.)

density

2.32 g/mL at 25 °C (lit.)

application(s)

cell analysis

SMILES string

Cl[Mg]Cl

InChI

1S/2ClH.Mg/h2*1H;/q;;+2/p-2

InChI key

TWRXJAOTZQYOKJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium chloride is a white crystalline solid which is soluble in water and alcohol. It has cubic close packing structure with alternate layers of octahedral holes occupied by Mg2+ ions. It is widely used in the field of solar cells and batteries.

Application

Magnesium chloride can be used:
  • To fabricate passivated ZnO nanoparticles as electron transport layer for PbS quantum dot solar cells. This will reduce the cell defects and significantly improves efficiency.
  • To prepare dual salt electrolyte for hydrid Mg/Li rechargeable batteries to enhance ionic conductivity.
  • As a dopant to prepare Mg doped TiO2 electron transport layer for low-temperature processed planar heterojunction perovskite solar cells. Mg doping significantly helps to enhance the power conversion efficiency of solar cells.

Caution

The appearance of gray/black specks should not affect use.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of temperature and humidity on electrospinning.
J. Mater. Sci., 44(5), 1357-1362 (2009)
Manal A Farg et al.
Human molecular genetics, 23(13), 3579-3595 (2014-02-20)
Intronic expansion of a hexanucleotide GGGGCC repeat in the chromosome 9 open reading frame 72 (C9ORF72) gene is the major cause of familial amyotrophic lateral sclerosis (ALS) and frontotemporal dementia. However, the cellular function of the C9ORF72 protein remains unknown.
Benjamin Dombert et al.
PloS one, 9(10), e110846-e110846 (2014-10-23)
Spinal muscular atrophy (SMA) is caused by deficiency of the ubiquitously expressed survival motoneuron (SMN) protein. SMN is crucial component of a complex for the assembly of spliceosomal small nuclear ribonucleoprotein (snRNP) particles. Other cellular functions of SMN are less
Fernando Cartón-García et al.
Scientific reports, 5, 12312-12312 (2015-07-24)
Inherited MYO5B mutations have recently been associated with microvillus inclusion disease (MVID), an autosomal recessive syndrome characterized by intractable, life-threatening, watery diarrhea appearing shortly after birth. Characterization of the molecular mechanisms underlying this disease and development of novel therapeutic approaches
Kadri Õunap et al.
PloS one, 10(7), e0133841-e0133841 (2015-07-28)
The human WBSCR22 protein is a 18S rRNA methyltransferase involved in pre-rRNA processing and ribosome 40S subunit biogenesis. Recent studies have shown that the protein function in ribosome synthesis is independent of its enzymatic activity. In this work, we have

Questions

1–7 of 7 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How do I store MgCl2 solution, once it's dissolved in water?

    1 answer
    1. When filter sterilized, this product may be stored at room temperature for up to 2 years, provided no contaminants are introduced. Please see the link below to review the product datasheet for item M1028, a prepared 1M solution:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/206/954/m1028pis.pdf

      Helpful?

  4. Would it be suitable for biochemical applications, such as preparing a buffer solution for bacterial treatment? Or would it be preferable to order a ready-to-use 1M solution (ex. M1028)?

    1 answer
    1. Product M8266 can be used in buffer preparation. The salt can be selected if you require a solution at a specific concentration that is not commercially available. This would require you to also have access to a suitable water source. If you decide to choose a solution this would reduce your preparation time. It is necessary to determine if a specific volume and concentration is required for the intended application.

      Helpful?

  5. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  6. What is the particle size of Product M8266, Magnesium chloride?

    1 answer
    1. According to our supplier, the particle size will range from - 10 + 100 mesh.

      Helpful?

  7. What is Product M8266, Magnesium chloride, soluble in?

    1 answer
    1. Per the chemicals encyclopedia published by the Royal Society of Chemistry, 13th Edition, this product is soluble in water (100 mg/ml), yielding a hazy, colorless solution.  Please also note that this could be a very exothermic reaction.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service