P2317
10X PCR Buffer without MgCl2
Optimized for routine PCR without MgCl2
Synonym(s):
Magnesium-free PCR buffer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106306
NACRES:
NA.52
Recommended Products
form
liquid
technique(s)
PCR: suitable
color
colorless
application(s)
agriculture
shipped in
wet ice
storage temp.
−20°C
Related Categories
General description
10X PCR Buffer II was tested at a final concentration of 1X (10mM Tris-HCl, pH 8.3 at 25 °C, 50mM KCl), in reactions containing 1-4mM MgCl2, each dNTP at 200 μM, primers defining an approximately 500 base pair region of λ DNA at 1.0μM each, λ DNA template at 1ng/100μL, and Taq DNA polymerase at 2.5 units/100μL. The reaction underwent 25 cycles of 94 °C to denature the double stranded DNA, 55 °C to anneal the DNA segments and 72 °C to extend the DNA segments. Following electrophoresis of the reaction products in 1.5% agarose gel, a single band of approximately 500 base pairs was visualized for PCRs containing 1-1.5mM MgCl2.
Application
10X PCR Buffer without MgCl2 has been used as a component of polymerase chain reaction (PCR) amplification mixture for the mycotoxigenic mould DNA, parasite DNA and Fusarium oxysporum DNA amplification.It has also been used as a component of the PCR mix for the amplification of nuclear ribosomal internal transcribed spacer (ITS2) and the partial ribulose-1,5 bisphosphate carboxylase/oxygenase large subunit rbcl gene for molecular and morphological characterization of Ulva sp from the Persian Gulf.
10× PCR Buffer without MgCl2 has been used with Sigma′s PCR enzymes.
Features and Benefits
- This product is tested for the absence of DNase and RNase.
- Suitable for use with magnesium chloride.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A simple method of preparing plant samples for PCR.
H Wang et al.
Nucleic acids research, 21(17), 4153-4154 (1993-08-25)
Andrée-Anne Dussault et al.
Biological procedures online, 8, 1-10 (2006-02-01)
Real-time polymerase chain reaction (PCR) constitutes a significant improvement over traditional end-point PCR, as it allows the quantification of starting amounts of nucleic acid templates, in real-time. However, quantification requires validation through numerous internal controls and standard curves. We describe
Molecular characterization of Fusarium oxysporum f. sp. cubense isolates from banana
Das A, et al.
Pest Management Science, 18(2), 171-178 (2012)
Mould incidence and mycotoxin contamination in freshly harvested maize kernels originated from India
Mudili V, et al.
Journal of the Science of Food and Agriculture, 94(13), 2674-2683 (2014)
Avoiding false positives with PCR.
Kwok, S., and Higuchi, R.
Nature, 229, 237-238 (1989)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service