D7295
Deoxynucleotide Mix, 10 mM
Molecular Biology Reagent
Synonym(s):
Deoxynucleotide Mix, 10 mM, 10mM dNTP mix, dNTP mix, dNTPs
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106305
NACRES:
NA.52
Recommended Products
Quality Level
form
liquid
concentration
10 mM
color
colorless
application(s)
agriculture
foreign activity
DNase, RNase, none detected
shipped in
dry ice
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Nucleotides are organic molecules that serve as the monomers, or subunits, of nucleic acids (like DNA and RNA). The building blocks of nucleic acids, nucleotides consist of a nitrogenous base (purine or pyrimidine), a five-carbon sugar (ribose or deoxyribose), and at least one phosphate group. A nucleoside along with a phosphate group yields a nucleotide. The Deoxynucleotide mix is a convenient premixed dNTP solution containing 10 mM each of UltraPure dATP, dCTP, dGTP, and TTP sodium salts in high-quality molecular biology grade water. One µL is sufficient for a standard 50 µL PCR reaction.
Application
dNTP Mix has been used:
- in the PCR amplification of genomic DNA isolated from insect, fungi, virus, human,
- in reverse transcription of total RNA to cDNA.
- as a component of the DNA amplification mixture for polymerase chain reaction (PCR)
- routine and long PCR
- manual and automated DNA sequencing
- cDNA synthesis and labeling reactions
Features and Benefits
- Purity of each dNTP: Minimum 99%
- Conveniently formulated; 1 μL is used per 50 μL PCR
- Equimolar amounts of each dNTP means less pipetting
- Minimize risk of contamination in PCR
- UltraPure dNTPs can help maximize consistency and yields in critical PCR reactions
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
The Field Cricket Gryllus assimilis and Two New Sister Species (Orthoptera: Gryllidae)
Weissman, et al.
Annals of the Entomological Society of America, 102, 367-380 (2009)
Billions and billions sold: Pet-feeder crickets (Orthoptera: Gryllidae), commercial cricket farms, an epizootic densovirus, and government regulations make for a potential disaster
Weissman, et al.
Zootaxa, 3504(1), 67-88 (2012)
Unal Egeli et al.
Cancer investigation, 24(5), 484-491 (2006-08-31)
BRCA1 and BRCA2 gene mutations in patients with breast and/or ovarian cancer have been not characterized in the Turkish population until now. A total of 87 female subjects from two sets of families (38 families total) provided blood samples from
Ajay V Maker et al.
Journal of the American College of Surgeons, 228(5), 721-729 (2019-02-23)
Current standard-of-care technologies, such as imaging and cyst fluid analysis, are unable to consistently distinguish intraductal papillary mucinous neoplasms (IPMNs) of the pancreas at high risk of pancreatic cancer from low-risk IPMNs. The objective was to create a single-platform assay
Sambrook, J., et al.
Molecular Cloning: A Laboratory Manual, 8-8 (1989)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service