Skip to Content
Merck
All Photos(1)

Documents

M8787

Sigma-Aldrich

Magnesium chloride solution

PCR Reagent, 25 mM MgCI2 solution for PCR

Synonym(s):

PCR optimization

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCl2
CAS Number:
Molecular Weight:
95.21
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.52

grade

PCR Reagent

Quality Level

200
300

form

liquid

packaging

vial of 1.5 mL

concentration

25 mM±1 mM

technique(s)

PCR: suitable

color

colorless

application(s)

agriculture
diagnostic assay manufacturing

foreign activity

DNase, RNase, none detected

storage temp.

2-8°C

SMILES string

Cl[Mg]Cl

InChI

1S/2ClH.Mg/h2*1H;/q;;+2/p-2

InChI key

TWRXJAOTZQYOKJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium chloride solution is suitable for the optimization of polymerase chain reactions (PCR). Magnesium chloride acts as a source of magnesium ions for polymerase chain reactions (PCR). It influences the primer-template annealing temperature, fidelity, specificity, and yield. Excess amounts of magnesium can cause an increase in nonspecific products, while lack of magnesium can lead to reduced yield.

Application

Magnesium chloride solution has been used as a component of the:

  • amplification mixture for polymerase chain reaction (PCR)
  • amplification mixture in an inter simple sequence repeats (ISSR)-PCR
  • reaction buffer in 16S-23S rRNA gene internal transcribed spacer (ITS) PCR amplification and restriction analysis (ITS-PCR-RFLP)
  • PCR master mix for quantitative reverse transcription-polymerase chain reaction (qRT PCR) to amplify reversely transcribed cDNA
  • reaction mix for RT-PCR to assess the gene expression

Suitability

Suitable for optimization of polymerase chain reactions.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular characterization of Fusarium oxysporum f. sp. cubense isolates from banana
Das A, et al.
Pest Management in Horticultural Ecosystems , 18(2), 171-178 (2012)
Massimiliano Bergallo et al.
American journal of perinatology, 36(10), 1060-1065 (2018-12-01)
Transcription of human endogenous retrovirus (HERV) elements is usually suppressed by epigenetic factors such as DNA methylation and heterochromatin silencing by histone modifications. There is an association between maternal smoking during pregnancy and DNA methylation levels in placental tissue and
Genome subtyping of autochthonous Bacillus species isolated from Iru, a fermented Parkia biglobosa seed
Adewumi G A, et al.
Food Biotechnology, 28(3), 250-268 (2014)
Kadri Õunap et al.
PloS one, 10(7), e0133841-e0133841 (2015-07-28)
The human WBSCR22 protein is a 18S rRNA methyltransferase involved in pre-rRNA processing and ribosome 40S subunit biogenesis. Recent studies have shown that the protein function in ribosome synthesis is independent of its enzymatic activity. In this work, we have
Takeshi Tomita et al.
PloS one, 9(9), e108957-e108957 (2014-10-01)
Celastramycin A, a small molecule that inhibits the production of antibacterial peptides in an ex vivo culture system of Drosophila, suppresses the TNFα-mediated induction of IL-8 in mammalian cells. To understand its molecular mechanism, we examined Celastramycin A binding proteins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service