Skip to Content
Merck
All Photos(1)

Documents

N5536

Sigma-Aldrich

Nefazodone hydrochloride

≥98% (HPLC), solid, antidepressant

Synonym(s):

2-[3-[4-(3-chlorophenyl)-1-piperazinyl]propyl]-5-ethyl- 2,4-dihydro-4-(2-phenoxyethyl)-3H-1,2,4-triazol-3-one hydrochloride, BMY-13754, MJ-13754-1, Serzone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H32ClN5O2 · HCl
CAS Number:
Molecular Weight:
506.47
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Nefazodone hydrochloride, ≥98% (HPLC), solid

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

white

solubility

DMSO: ≥10 mg/mL
H2O: insoluble

originator

Bristol-Myers Squibb

SMILES string

O=C1N(CCCN2CCN(C3=CC=CC(Cl)=C3)CC2)N=C(CC)N1CCOC4=CC=CC=C4.Cl

InChI

1S/C25H32ClN5O2.ClH/c1-2-24-27-31(25(32)30(24)18-19-33-23-10-4-3-5-11-23)13-7-12-28-14-16-29(17-15-28)22-9-6-8-21(26)20-22;/h3-6,8-11,20H,2,7,12-19H2,1H3;1H

InChI key

DYCKFEBIOUQECE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Novel antidepressant; mixed 5-HT2A serotonin receptor antagonist/serotonin uptake inhibitor.

Features and Benefits

This compound was developed by Bristol-Myers Squibb. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S C Collins et al.
Diabetologia, 51(9), 1689-1693 (2008-07-16)
Long-term exposure to NEFAs leads to inhibition of glucose-induced insulin secretion. We tested whether the release of somatostatin and glucagon, the two other major islet hormones, is also affected. Mouse pancreatic islets were cultured for 72 h at 4.5 or
Jonathan N Bauman et al.
Drug metabolism and disposition: the biological fate of chemicals, 36(6), 1016-1029 (2008-03-12)
In vitro metabolism/bioactivation of structurally related central nervous system agents nefazodone (hepatotoxin) and aripiprazole (nonhepatotoxin) were undertaken in human liver microsomes in an attempt to understand the differences in toxicological profile. NADPH-supplemented microsomal incubations of nefazodone and glutathione generated conjugates
James A Dykens et al.
Toxicological sciences : an official journal of the Society of Toxicology, 103(2), 335-345 (2008-03-18)
Mitochondrial toxicity is increasingly implicated in a host of drug-induced organ toxicities, including hepatotoxicity. Nefazodone was withdrawn from the U.S. market in 2004 due to hepatotoxicity. Accordingly, we evaluated nefazodone, another triazolopyridine trazodone, plus the azaspirodecanedione buspirone, for cytotoxicity and
Rachel E Maddux et al.
Journal of affective disorders, 117(3), 174-179 (2009-02-17)
Individuals with chronic depression respond poorly to both medication and psychotherapy. The reasons for the poorer response, however, remain unclear. One potential factor is the presence of comorbid Axis II personality disorders (PDs), which occur at high rates among these
Nader Perroud et al.
Pharmacogenomics, 12(3), 365-377 (2011-04-01)
This study investigated clinical and genetic predictors of increasing suicidal ideation during antidepressant treatment. A total of 131 depressed outpatients were allocated to four antidepressants (paroxetine, venlafaxine, clomipramine or nefazodone) in a sequential step procedure until remission. Suicidality was assessed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service