Skip to Content
Merck

Skip To

B7148

Buspirone hydrochloride

≥99% (TLC), powder, 5-HT1A serotonin receptor agonist

Synonym(s):

N-[4-[4-(2-Pyrimidinyl)-1-piperazinyl]butyl]-8-azaspiro[4.5]decane-7,9-dione hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H31N5O2 · HCl
CAS Number:
Molecular Weight:
421.96
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
251-489-4
MDL number:
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Buspirone hydrochloride,

form

powder

Quality Segment

originator

Bristol-Myers Squibb

storage temp.

2-8°C

SMILES string

Cl[H].O=C1CC2(CCCC2)CC(=O)N1CCCCN3CCN(CC3)c4ncccn4

InChI

1S/C21H31N5O2.ClH/c27-18-16-21(6-1-2-7-21)17-19(28)26(18)11-4-3-10-24-12-14-25(15-13-24)20-22-8-5-9-23-20;/h5,8-9H,1-4,6-7,10-17H2;1H

InChI key

RICLFGYGYQXUFH-UHFFFAOYSA-N

Gene Information

human ... HTR1A(3350)

Biochem/physiol Actions

5-HT1A serotonin receptor agonist; anxiolytic.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Bristol-Myers Squibb. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR19171078802Y0000131
form

powder

form

solid

form

-

form

-

Quality Level

100

Quality Level

300

Quality Level

-

Quality Level

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

originator

Bristol-Myers Squibb

originator

-

originator

-

originator

-

Gene Information

human ... HTR1A(3350)

Gene Information

human ... HTR1A(3350)

Gene Information

human ... HTR1A(3350)

Gene Information

human ... HTR1A(3350)


Still not finding the right product?

Explore all of our products under Buspirone hydrochloride


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters