Skip to Content
Merck
All Photos(3)

Key Documents

A3035

Sigma-Aldrich

Acetaminophen

analytical standard

Synonym(s):

4′-Hydroxyacetanilide, 4-Acetamidophenol, N-(4-Hydroxyphenyl)acetamide, N-Acetyl-4-aminophenol, APAP, Paracetamol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC6H4OH
CAS Number:
Molecular Weight:
151.16
Beilstein:
2208089
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

form

powder

packaging

vial of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

168-172 °C (lit.)

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

SMILES string

CC(=O)Nc1ccc(O)cc1

InChI

1S/C8H9NO2/c1-6(10)9-7-2-4-8(11)5-3-7/h2-5,11H,1H3,(H,9,10)

InChI key

RZVAJINKPMORJF-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Analgesic.

Packaging

Supplied in amber screw-cap vials

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

364.3 °F - Pensky-Martens closed cup

Flash Point(C)

184.6 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Garry G Graham et al.
Inflammopharmacology, 21(3), 201-232 (2013-05-31)
Paracetamol is used worldwide for its analgesic and antipyretic actions. It has a spectrum of action similar to that of NSAIDs and resembles particularly the COX-2 selective inhibitors. Paracetamol is, on average, a weaker analgesic than NSAIDs or COX-2 selective
Garry G Graham et al.
Drug safety, 28(3), 227-240 (2005-03-01)
The excellent tolerability of therapeutic doses of paracetamol (acetaminophen) is a major factor in the very wide use of the drug. The major problem in the use of paracetamol is its hepatotoxicity after an overdose. Hepatotoxicity has also been reported
Mitchell R McGill et al.
Pharmaceutical research, 30(9), 2174-2187 (2013-03-07)
Acetaminophen (APAP) is one of the most widely used drugs. Though safe at therapeutic doses, overdose causes mitochondrial dysfunction and centrilobular necrosis in the liver. The first studies of APAP metabolism and activation were published more than 40 years ago.
Karel Allegaert et al.
Archives of disease in childhood, 98(6), 462-466 (2013-04-23)
There remains a need for alternative medical treatments for patent ductus arteriosus (PDA) closure in extreme preterm neonates because of therapeutic failure and adverse effects associated with non-selective cyclo-oxygenase inhibitors. Reports of an association between paracetamol exposure and PDA closure
S M de Morais et al.
Gastroenterology, 102(2), 577-586 (1992-02-01)
Gilbert's syndrome occurs in 5%-7% of the human population and is caused by an inherited deficiency in the glucuronidation of endogenous bilirubin, resulting in its accumulation and jaundice. The authors of the present study have previously shown that rats with

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service