Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C13H18ClNO · HCl
CAS Number:
Molecular Weight:
276.20
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
250-759-9
MDL number:
Product Name
Bupropion hydrochloride, ≥98% (HPLC), solid
description
Drug Control: Kábítószer / Narcotic Drug (Hungary), 78/2022. (XII. 28.) BM rendelet
Quality Level
assay
≥98% (HPLC)
form
solid
drug control
Kábítószer / Narcotic Drug (Hungary), 78/2022. (XII. 28.) BM rendelet
color
white
solubility
H2O: 15 mg/mL, clear
originator
GlaxoSmithKline
SMILES string
Cl[H].CC(NC(C)(C)C)C(=O)c1cccc(Cl)c1
InChI
1S/C13H18ClNO.ClH/c1-9(15-13(2,3)4)12(16)10-6-5-7-11(14)8-10;/h5-9,15H,1-4H3;1H
InChI key
HEYVINCGKDONRU-UHFFFAOYSA-N
Gene Information
human ... DRD1(1812), DRD2(1813), DRD3(1814), DRD4(1815), DRD5(1816), SLC6A2(6530), SLC6A3(6531)
General description
Bupropion is structurally linked to the phenylethylamine family. It is a monocyclic aminoketone (chloropropiophenone).
Application
Bupropion hydrochloride has been used as an anti-depressant and injected into mice intraperitoneally for forced swim test. It has also been used to study its effects on depression, anxiety and smoking cessation.
Biochem/physiol Actions
Inhibits the dopamine and norepinephrine transporters with Kis of 2.8 μM and 1.4 μM, respectively. Does not inhibit the serotonin transporter (Ki = 45 μM). Antidepressant.
Bupropion possesses antidepressant and anxiolytic properties. It serves as a good auxiliary for people who makes an effort to give up smoking. It may also be used in the treatment for attention-deficit hyperactivity disorder.
Features and Benefits
This compound is a featured product for ADME Tox and Neuroscience research. Discover more featured ADME Tox and Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.
Still not finding the right product?
Explore all of our products under Bupropion hydrochloride
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Effects of bupropion on depression, anxiety and smoking cessation
Hernandez-Lozano M, et al.
Archives of Neuroscience, 15(4), 260-266 (2010)
B R Cooper et al.
The Journal of pharmacology and experimental therapeutics, 215(1), 127-134 (1980-10-01)
Bupropion (BW 323U; Wellbutrin), a novel compound with antidepressant effects in man, was found to reduce immobility in an "experimental helplessness" forced swimming antidepressant test in rats as did imipramine and amitriptyline. Higher doses produced elevated locomotor activity in an
Jinhui Wang et al.
Frontiers in pharmacology, 11, 593518-593518 (2021-03-23)
Poziotinib is an orally active, irreversible, pan-HER tyrosine kinase inhibitor used to treat non-small cell lung cancer, breast cancer, and gastric cancer. Poziotinib is currently under clinical investigation, and understanding its drug-drug interactions is extremely important for its future development
Global Trade Item Number
| SKU | GTIN |
|---|---|
| B102-50MG | 04061833420157 |
| B102-250MG | 04061833420133 |
| B102-1G | 04061825980478 |
