Skip to Content
Merck
All Photos(3)

Key Documents

F8384

Sigma-Aldrich

Flavin adenine dinucleotide disodium salt hydrate

BioReagent, suitable for cell culture

Synonym(s):

FAD, FAD-Na2, Riboflavin 5′-adenosine diphosphate disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H31N9Na2O15P2 · xH2O
CAS Number:
Molecular Weight:
829.51 (anhydrous basis)
Beilstein:
5326842
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

biological source

bovine milk

product line

BioReagent

Assay

≥95%

form

powder

technique(s)

cell culture | mammalian: suitable

solubility

water: 50 mg/mL, clear

storage temp.

−20°C

SMILES string

[Na+].[Na+].[H]O[H].Cc1cc2N=C3C(=O)NC(=O)N=C3N(C[C@H](O)[C@H](O)[C@H](O)COP([O-])(=O)OP([O-])(=O)OC[C@H]4O[C@H]([C@H](O)[C@@H]4O)n5cnc6c(N)ncnc56)c2cc1C

InChI

1S/C27H33N9O15P2.2Na.H2O/c1-10-3-12-13(4-11(10)2)35(24-18(32-12)25(42)34-27(43)33-24)5-14(37)19(39)15(38)6-48-52(44,45)51-53(46,47)49-7-16-20(40)21(41)26(50-16)36-9-31-17-22(28)29-8-30-23(17)36;;;/h3-4,8-9,14-16,19-21,26,37-41H,5-7H2,1-2H3,(H,44,45)(H,46,47)(H2,28,29,30)(H,34,42,43);;;1H2/q;2*+1;/p-2/t14-,15+,16+,19-,20+,21+,26+;;;/m0.../s1

InChI key

GXTPHHZYFMAGLX-UJXBNFGUSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Flavin adenine dinucleotide (FAD) is a redox cofactor of flavoproteins, such as cytochrome-b5 reductase enzyme. With the help of nicotinamide adenine dinucleotide phosphate (NADPH), FAD can be reduced to form FADH2.

Application

Flavin adenine dinucleotide (FAD) is used as a redox cofactor (electron carrier) by flavoproteins including succinate dehydrogenase (complex), α-ketoglutarate dehydrogenase, apoptosis-inducing factor 2 (AIF-M2, AMID), folate/FAD-dependent tRNA methyltransferases, and N-hydroxylating flavoprotein monooxygenases. FAD is a component of the pyruvate dehydrogenase complex.
Flavin adenine dinucleotide disodium salt hydrate has been used as a component in the extraction buffer for plant tissue extraction. It has also been used to study its effects on infarct areas of middle cerebral artery occlusion (MCAO) induced mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bone Marrow, Blood Cells, and the Lymphoid/Lymphatic System1
Pathologic Basis of Veterinary Disease, 724-804 (2017)
Mechanisms of Hepatocyte Detoxification
Physiology of the Gastrointestinal Tract, 2, 1507-1527 (2012)
In vitro Nitrate Reductase Activity Assay from Arabidopsis Crude Extracts
Kim JY and Seo HS
Bio-protocol, 8(7) (2018)
Ying-Xin Zou et al.
CNS neuroscience & therapeutics, 18(10), 834-840 (2012-08-29)
To explore risk factors for stroke independent of hypertension and the relationship between riboflavin kinase (RFK) and stroke. Gene expression profiling in the brains of spontaneously hypertensive rats (SHR) and stroke-prone spontaneously hypertensive rats (SHRSP) was comparatively analyzed by gene
Saurav Roy Choudhury et al.
Cell, 178(5), 1205-1221 (2019-08-24)
A hallmark feature of inflammation is the orchestrated recruitment of neutrophils from the bloodstream into inflamed tissue. Although selectins and integrins mediate recruitment in many tissues, they have a minimal role in the lungs and liver. Exploiting an unbiased in vivo

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service