Skip to Content
Merck
All Photos(1)

Documents

N1915

Sigma-Aldrich

Nebivolol hydrochloride

≥98% (HPLC)

Synonym(s):

Narbivolol, Nebilet, R-65824, rel-(aR,a′R,2R,2′S)-a,a′-[iminobis(methylene)]bis[6-fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol] hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H25F2NO4 · HCl
CAS Number:
Molecular Weight:
441.90
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

DMSO: ≥20 mg/mL

originator

Forest Labs

storage temp.

room temp

SMILES string

Cl.O[C@@H](CNC[C@H](O)[C@@H]1CCc2cc(F)ccc2O1)[C@H]3CCc4cc(F)ccc4O3

InChI

1S/C22H25F2NO4.ClH/c23-15-3-7-19-13(9-15)1-5-21(28-19)17(26)11-25-12-18(27)22-6-2-14-10-16(24)4-8-20(14)29-22;/h3-4,7-10,17-18,21-22,25-27H,1-2,5-6,11-12H2;1H/t17-,18-,21-,22+;/m0./s1

InChI key

JWEXHQAEWHKGCW-BIISKSHESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nebivolol hydrochloride has been used as a standard in the validation of high-performance thin-layer chromatography (HPTLC)-densitometry method. It may be used in calibration curve preparation and absorption spectrum studies using spectrophotometry.

Biochem/physiol Actions

Nebivolol hydrochloride (NEB) is used as a racemic mixture for clinical studies. It stabilizes membrane and possesses intrinsic sympathomimetic functionality. Nebivolol interaction with π-acceptors (2,3-dichloro-5,6-dicyano-1,4-benzoquinone (DDQ), 2,4-dinitrophenol (DNP) and 2,4,6-trinitrophenol (picric acid; PA)) is useful in the spectrophotometric detection methods.
Nebivolol is a competitive, highly selective b1-receptor antagonist with mild vasodialating properties, possibly due to an interaction with the L-arginine/ NO pathway, and is used for treatment of essential hypertension. It is 40-fold selective for β1 vs. β2, and lacks ISA (intrinsic sympathomimetic activity).

Features and Benefits

This compound is featured on the β-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Forest Labs. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vijay Singh Jatav et al.
Advanced biomedical research, 2, 62-62 (2013-11-14)
Nebivolol hydrochloride is a third generation β-blocker with highly selective β1-receptor antagonist with antihypertensive properties having plasma half life of 10 h and 12% oral bioavailability. The aim of the present investigation was to form matrix type transdermal patches containing
New spectrophotometric determination of nebivolol in pharmaceutical formulation via charge transfer complex
Alhemiary, Nabil AF
Austin Journal of Analytical and Pharmaceutical Chemistry , 4(2), 1084-1091 (2017)
Development of quantitative HPTLC-Densitometry methods for the analysis of amiodarone HCl, carvedilol, doxylamine succinate, magnesium salicylate, metoprolol succinate, nebivolol HCl, and salicylamide using a model process developed earlier for the transfer of TLC screening methods
Nguyen K and Sherma J
Acta Chromatographica , 30(4), 264-268 (2018)
Deepak Sharma et al.
Pharmaceutical methods, 2(1), 9-14 (2011-01-01)
The present study aimed to develop and validate the simultaneous estimation of amlodipine and nebivolol in tablet dosage forms. An isocratic reversed phase high-performance liquid chromatographic (HPLC) method with ultraviolet detection at 268 nm has been developed for the determination
Nebivolol Hydrochloride Loaded Nanostructured Lipid Carriers as Transdermal Delivery System: Part 1: Preparation, Characterization and In Vitro Evaluation
Ghazy E, et al.
Al-Mustansiriyah Journal of Pharmaceutical Sciences (AJPS), 16(2), 14-Jan-14-Jan (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service