Skip to Content
Merck
All Photos(3)

Key Documents

D9815

Sigma-Aldrich

Doxazosin mesylate

≥97% (HPLC), powder

Synonym(s):

1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-[4-(1,4-benzodioxan-2-yl)carpiperazin-1-yl)]-6,7-dimethoxyquinazoline mesylate, Doxazosin methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25N5O5 · CH3SO3H
CAS Number:
Molecular Weight:
547.58
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

form

powder

color

white

SMILES string

CS(O)(=O)=O.COc1cc2nc(nc(N)c2cc1OC)N3CCN(CC3)C(=O)C4COc5ccccc5O4

InChI

1S/C23H25N5O5.CH4O3S/c1-30-18-11-14-15(12-19(18)31-2)25-23(26-21(14)24)28-9-7-27(8-10-28)22(29)20-13-32-16-5-3-4-6-17(16)33-20;1-5(2,3)4/h3-6,11-12,20H,7-10,13H2,1-2H3,(H2,24,25,26);1H3,(H,2,3,4)

InChI key

VJECBOKJABCYMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

α1-adrenoceptor antagonist; relaxes smooth muscles of the prostate

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sakip Erturhan et al.
Urology, 81(3), 640-643 (2013-01-25)
To evaluate the possible efficacy of an α1 blocker (doxazosin) therapy on the management and disease course in pediatric patients with distal ureteral stones. A total of 45 patients (24 boys and 21 girls) with a single lower ureteral stone
Cédric M Hysek et al.
Psychopharmacology, 224(3), 363-376 (2012-06-16)
Pupillometry can be used to characterize autonomic drug effects. This study was conducted to determine the autonomic effects of 3,4-methylenedioxymethamphetamine (MDMA, ecstasy), administered alone and after pretreatment with reboxetine, duloxetine, clonidine, carvedilol, and doxazosin, on pupillary function. Infrared pupillometry was
Aaron Petty et al.
PloS one, 7(8), e42120-e42120 (2012-08-24)
During tumor progression, EphA2 receptor can gain ligand-independent pro-oncogenic functions due to Akt activation and reduced ephrin-A ligand engagement. The effects can be reversed by ligand stimulation, which triggers the intrinsic tumor suppressive signaling pathways of EphA2 including inhibition of
Giovanni Conzo et al.
International journal of surgery (London, England), 11(2), 152-156 (2012-12-27)
Minimally invasive adrenalectomy for pheochromocytoma (PCC) is a complex surgical procedure especially because of the haemodynamic instability due to the excessive secretion of cathecolamines, which may result in a considerable risk of conversion and complications. By a multicentric retrospective study
Thomas F Newton et al.
PloS one, 7(2), e30854-e30854 (2012-02-10)
Preclinical research implicates dopaminergic and noradrenergic mechanisms in mediating the reinforcing effects of drugs of abuse, including cocaine. The objective of this study was to evaluate the impact of treatment with the noradrenergic α(1) receptor antagonist doxazosin on the positive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service