Skip to Content
Merck
All Photos(1)

Key Documents

M1830000

Metoprolol tartrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±)-Metoprolol (+)-tartrate salt, (±)1-(Isopropylamino)-3-[p-(β-methoxyethyl)phenoxy]-2-propanol (+)-tartrate salt, Lopressor

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C15H25NO3)2 · C4H6O6
CAS Number:
Molecular Weight:
684.81
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

metoprolol

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical

format

neat

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O.COCCc1ccc(OCC(O)CNC(C)C)cc1.COCCc2ccc(OCC(O)CNC(C)C)cc2

InChI

1S/2C15H25NO3.C4H6O6/c2*1-12(2)16-10-14(17)11-19-15-6-4-13(5-7-15)8-9-18-3;5-1(3(7)8)2(6)4(9)10/h2*4-7,12,14,16-17H,8-11H2,1-3H3;1-2,5-6H,(H,7,8)(H,9,10)/t;;1-,2-/m..1/s1

InChI key

YGULWPYYGQCFMP-CEAXSRTFSA-N

Gene Information

human ... ADRB1(153)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jieqiong Zhao et al.
Cell & bioscience, 10, 72-72 (2020-06-04)
Arginine vasopressin (AVP) is elevated in patients with heart failure, and the increase in the AVP concentration in plasma is positively correlated with disease severity and mortality. Metoprolol (Met) is a beta blocker that is widely used in the clinic
Martin H Ruwald et al.
Journal of the American College of Cardiology, 61(14), 1518-1526 (2013-03-19)
This study sought to compare the effects of metoprolol and carvedilol in the MADIT-CRT (Multicenter Automatic Defibrillator Implantation Trial With Cardiac Resynchronization Therapy) study. The impact of beta-blockers in heart failure (HF) patients with devices is uninvestigated. All patients receiving
C M Blake et al.
Clinical pharmacology and therapeutics, 94(3), 394-399 (2013-05-15)
Metoprolol, a commonly prescribed β-blocker, is primarily metabolized by cytochrome P450 2D6 (CYP2D6), an enzyme with substantial genetic heterogeneity. Several smaller studies have shown that metoprolol pharmacokinetics is influenced by CYP2D6 genotype and metabolizer phenotype. To increase robustness of metoprolol
Thomas Kahan et al.
European journal of clinical investigation, 43(2), 141-151 (2013-01-03)
Although stable angina pectoris often carries a favourable prognosis, it remains important to identify patients with an increased risk of cardiovascular (CV) complications. Many new markers of disease activity and prognosis have been described. We evaluated whether common and easily
Mehmet Ozaydin et al.
European heart journal, 34(8), 597-604 (2012-12-13)
Carvedilol and N-acetyl cysteine (NAC) have antioxidant and anti-inflammatory properties. Aim was to evaluate the efficacy of metoprolol, carvedilol, and carvedilol plus NAC on the prevention of post-operative atrial fibrillation (POAF). Patients undergoing cardiac surgery (n = 311) were randomized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service