Skip to Content
Merck
All Photos(3)

Key Documents

R0875

Sigma-Aldrich

Reserpine

Synonym(s):

(3β, 16β, 17α, 18β, 20α)-11,17-Dimethoxy-18-[(3,4,5-trimethoxybenzoyl)oxy]yohimban-16-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H40N2O9
CAS Number:
Molecular Weight:
608.68
Beilstein:
102014
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

mp

~265 °C (dec.)

SMILES string

CO[C@H]1[C@@H](C[C@@H]2CN3CCc4c([nH]c5cc(OC)ccc45)[C@H]3C[C@@H]2[C@@H]1C(=O)OC)OC(=O)c6cc(OC)c(OC)c(OC)c6

InChI

1S/C33H40N2O9/c1-38-19-7-8-20-21-9-10-35-16-18-13-27(44-32(36)17-11-25(39-2)30(41-4)26(12-17)40-3)31(42-5)28(33(37)43-6)22(18)15-24(35)29(21)34-23(20)14-19/h7-8,11-12,14,18,22,24,27-28,31,34H,9-10,13,15-16H2,1-6H3/t18-,22+,24-,27-,28+,31+/m1/s1

InChI key

QEVHRUUCFGRFIF-MDEJGZGSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reserpine has been used:
  • to study its effect on minimal inhibitory concentration of trovafloxacin and ciprofloxacin
  • inject rats, to study its effect on behavioural parameters such as locomotion and fear/anxiety
  • as a lock spray reference compound in lipid profiling

Biochem/physiol Actions

Inhibits vesicular uptake of catecholamines and serotonin.
Reserpine is an alkaloid present in Rauwolfia species. It is useful in treating systemic hypertension, schizophrenia, psychiatric disorders and Raynaud′s syndrome. Reserpine is known to inhibit adrenaline induced-cutaneous blood vessel constriction. It diminishes the effect of hyperthyroidism on cardiovascular and nervous system.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Repr. 1A - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Current Cardiovascular Drugs, 302-302 (2005)
Comparative effects of Rauwolfia vomitoria and chlorpromazine on locomotor behaviour and anxiety in mice
Bisong SA, et al.
Journal of Ethnopharmacology, 132(1), 334-339 (2010)
Effects of long-term intake of lactotripeptides on cardiovascular risk factors in hypertensive subjects
Jauhiainen T, et al.
European Journal of Clinical Nutrition, 66(7), 843-843 (2012)
In vitro activities of six quinolones and mechanisms of resistance in Staphylococcus aureus and coagulase-negative staphylococci
Linde H, et al.
Antimicrobial Agents and Chemotherapy, 45(5), 1553-1557 (2001)
Alexander C Sutton et al.
Journal of neurophysiology, 109(2), 363-374 (2012-10-19)
Deep brain stimulation (DBS) employing high-frequency stimulation (HFS) is commonly used in the globus pallidus interna (GPi) and the subthalamic nucleus (STN) for treating motor symptoms of patients with Parkinson's disease (PD). Although DBS improves motor function in most PD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service