Skip to Content
Merck
All Photos(2)

Key Documents

I127

Sigma-Aldrich

ICI 118,551 hydrochloride

≥98% (HPLC), powder, β2 inverse agonist

Synonym(s):

(2R,3R)-rel-3-isopropylamino-1-(7-methylindan-4-yloxy)-butan-2-ol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H27NO2 · HCl
CAS Number:
Molecular Weight:
313.86
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

ICI 118,551 hydrochloride, ≥98% (HPLC), powder

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white

solubility

H2O: >10 mg/mL

SMILES string

Cl.CC(C)N[C@@H](C)[C@@H](O)COc1ccc(C)c2CCCc12

InChI

1S/C17H27NO2.ClH/c1-11(2)18-13(4)16(19)10-20-17-9-8-12(3)14-6-5-7-15(14)17;/h8-9,11,13,16,18-19H,5-7,10H2,1-4H3;1H/t13-,16-;/m0./s1

InChI key

KBXMBGWSOLBOQM-LINSIKMZSA-N

Gene Information

human ... ADRB2(154)

Related Categories

Application

ICI 118,551 hydrochloride has been used:
  • to block β-adrenergic signaling in various experiments
  • to counteract degradation by endogenous T-lymphocyte monoamine oxidases
  • to determine the specific receptor subtype necessary for isoproterenol to induce up-regulation of gene transcripts

Biochem/physiol Actions

ICI 118,551 has been used to treat vasomotor migraines and somatic troubles related to anxiety state.
ICI 118,551 hydrochloride is a highly selective β2-adrenoceptor antagonist.

Features and Benefits

This compound is featured on the β-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of ICI 118551, a new beta2 blocking drug, and related compounds by RP-HPLC?DAD
Quaglia MG, et al.
Journal of Pharmaceutical and Biomedical Analysis, 10(10-12), 1081-1084 (1992)
beta-Adrenergic-stimulated macrophages: comprehensive localization in the M1-M2 spectrum
Lamkin DM, et al.
Brain, behavior, and immunity, 57(, 338-346 (2016)
R M Booze et al.
The Journal of pharmacology and experimental therapeutics, 249(3), 911-920 (1989-06-01)
The localization and number of beta adrenergic receptors were directly compared in the brains of rats and guinea pigs. The time course of association and saturability of [125I]cyanopindolol (CYP) binding to slide-mounted tissue sections was similar in rats (Kd =
Effect of chronic psychological stress on liver metastasis of colon cancer in mice
Zhao L, et al.
PLoS ONE, 10(10), e0139978-e0139978 (2015)
Hongliang Li et al.
The Journal of biological chemistry, 288(42), 30734-30741 (2013-09-18)
Functional autoantibodies to the autonomic receptors are increasingly recognized in the pathophysiology of cardiovascular diseases. To date, no human activating monoclonal autoantibodies to these receptors have been available. In this study, we describe for the first time a β2-adrenergic receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service