Skip to Content
Merck
All Photos(1)

Key Documents

M4642

Sigma-Aldrich

Minimum Essential Medium Eagle

With Hanks′ salts and ʟ-glutamine, without sodium bicarbonate, powder, suitable for cell culture

Synonym(s):

EMEM, MEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

Product Name

Minimum Essential Medium Eagle, With Hanks′ salts and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture

form

powder

technique(s)

cell culture | mammalian: suitable

components

glucose: yes
Hanks’ salts (2% CO2): yes
L-glutamine: yes
Earle’s salts (5% CO2): no
NaHCO3: no
phenol red: yes
sodium pyruvate: no
stable glutamine: no
HEPES: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Minimum Essential Medium Eagle (MEM) is a synthetic cell culture media, developed by Harry Eagle. It has high concentrations of amino acids, so the medium closely approximates the protein concentration of cultured mammalian cells. Optional supplementation of non-essential amino acids to the formulations that incorporate either Hanks′ or Earle′s salts has increased the applicability of this medium. This medium lacks calcium that permits the growth of cells in suspension. Modification of the original MEM formulation. Earle′s salts have been replaced with Hanks′ salts. Since the bicarbonate concentration is substantially lower, this medium has a substantially reduced buffering capacity.

Application

Minimum Essential Medium Eagle (MEM) has been used:

  • as a component of the Hank′s MEM (HMEM)-Pipes buffer to induce anaphylatoxin induced-ear swelling in mice
  • as a component of the slice culture medium (SCM) for the incubation of mouse hippocampal slice cultures
  • to culture Müller cells from guinea pigs

Quantity

Formulated to contain 10.6 grams of powder per liter of medium.

Reconstitution

Supplement with 0.35 g/L sodium bicarbonate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The activation of IL-8 receptors in cultured guinea pig Muller glial cells is modified by signals from retinal pigment epithelium
Iwona M G, et al.
Journal of Neuroimmunology, 161, 49-60 (2005)
Mast cell anaphylatoxin receptor expression can enhance IgE-dependent skin inflammation in mice
Beatrix S, et al.
The Journal of Allergy and Clinical Immunology, 131(2), 541-548 (2013)
Anthony Dudilot et al.
Current biology : CB, 30(7), 1167-1176 (2020-02-23)
Synaptic plasticity, with its two most studied forms, long-term potentiation (LTP) and long-term depression (LTD), is the cellular mechanism underlying learning and memory. Although it has been known for two decades that bidirectional synaptic plasticity necessitates a corresponding bidirectional regulation
John K Mich et al.
Cell reports, 34(13), 108754-108754 (2021-04-01)
Viral genetic tools that target specific brain cell types could transform basic neuroscience and targeted gene therapy. Here, we use comparative open chromatin analysis to identify thousands of human-neocortical-subclass-specific putative enhancers from across the genome to control gene expression in
Carlos Mendoza-Palomares et al.
Eukaryotic cell, 7(4), 684-697 (2008-02-19)
Cysteine proteases have been shown to be essential virulence factors and drug targets in trypanosomatids and an attractive antidisease vaccine candidate for Trypanosoma congolense. Here, we describe an important amplification of genes encoding cathepsin B-like proteases unique to T. congolense.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service